CLS70791N
Pyrex® serological pipettes, disposable, non-sterile, unplugged
volume 1.0 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
borosilicate glass
sterility
non-sterile
feature
DNase free: no
RNase free: no
graduations
subdivision
packaging
case of 1000 ea
pack of 250 ea
manufacturer/tradename
Corning 7079-1N
volume measuring range
1.0 mL
capacity
1 mL
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
- Disposable PYREX® glass pipettes are calibrated "to deliver" (TD) with blow-out
- Long, slender, tapered tips of pipettes make pipetting faster and easier
- All pipettes have negative graduations and fire polished tips for burr-free, even flow rates
- Nonsterile, unplugged and bulk packed.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service