Skip to Content
Merck
All Photos(1)

Key Documents

A54059

Sigma-Aldrich

2-(2-Aminoethoxy)ethanol

98%

Synonym(s):

Diethylene glycolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
105.14
Beilstein:
906728
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

bp

218-224 °C (lit.)

density

1.048 g/mL at 25 °C (lit.)

SMILES string

NCCOCCO

InChI

1S/C4H11NO2/c5-1-3-7-4-2-6/h6H,1-5H2

InChI key

GIAFURWZWWWBQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-(2-Aminoethoxy)ethanol is commonly used as a spacer/linker in the synthesis of bioconjugate materials for applications such as drug delivery and protein labeling. It is also employed as a receptor chain in the preparation of carboxamidoquinoline based water-soluble, ratiometric fluorescent zinc sensor.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

260.6 °F

Flash Point(C)

127 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Superresolution imaging of targeted proteins in fixed and living cells using photoactivatable organic fluorophores.
Lee, Hsiao-lu D et al.
Journal of the American Chemical Society, 132(43), 15099-15101 (2010)
Ratiometric and water-soluble fluorescent zinc sensor of carboxamidoquinoline with an alkoxyethylamino chain as receptor.
Zhang, Yu et al.
Organic Letters, 10(3), 473-476 (2008)
C Tournaire-Arellano et al.
Carbohydrate research, 314(1-2), 47-63 (1999-05-07)
Ureido and thioureido derivatives of 2-acetamido-2-deoxy-beta-D-glucose, 1-amino-1-deoxy-D-glucitol and 2-(2-aminoethoxy)ethanol were prepared as N-acetyl-beta-D-hexosaminidase (NAHase) inhibitors and were evaluated on Trichomonas vaginalis NAHase. Although none showed complete inhibition of the enzyme at 100 microM, 1-amino-1-deoxy-D-glucitol derivatives acted as competitive inhibitors of
Cao-An Vu et al.
ACS omega, 4(12), 14765-14771 (2019-09-26)
Silicon nanowire field-effect transistors (SiNW-FETs) have been demonstrated as a highly sensitive platform for label-free detection of a variety of biological and chemical entities. However, detecting signal from immunoassays by nano-FETs is severely hindered by the distribution of different charged
HaloTag: a novel protein labeling technology for cell imaging and protein analysis.
Los, Georgyi V et al.
ACS Chemical Biology, 3(6), 373-382 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service