Skip to Content
Merck
All Photos(1)

Key Documents

A37200

Sigma-Aldrich

Aminoacetaldehyde diethyl acetal

98%

Synonym(s):

2,2-Diethoxyethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH(OC2H5)2
CAS Number:
Molecular Weight:
133.19
Beilstein:
506067
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.417 (lit.)

bp

162-163 °C (lit.)

density

0.916 g/mL at 25 °C (lit.)

SMILES string

CCOC(CN)OCC

InChI

1S/C6H15NO2/c1-3-8-6(5-7)9-4-2/h6H,3-5,7H2,1-2H3

InChI key

HJKLEAOXCZIMPI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Su-Hwan Kim et al.
Biomaterials, 242, 119905-119905 (2020-03-08)
Epigallocatechin gallates (EGCGs), isolated from green tea, have intrinsic properties such as anti-oxidant, anti-inflammation, and radical scavenger effects. In this study, we report a tissue adhesive and anti-inflammatory hydrogel formed by high-affinity enzymatic crosslinking of polyphenolic EGCGs. A mixture of
Taehyung C Lee et al.
Cell chemical biology, 25(6), 797-801 (2018-04-24)
Single-molecule fluorescence is widely used to study conformational complexity in proteins, and has proven especially valuable with intrinsically disordered proteins (IDPs). Protein studies using dual-color single-molecule Förster resonance energy transfer (smFRET) are now quite common, but many could benefit from simultaneous
Hiraku Onishi et al.
Journal of drug targeting, 25(8), 747-753 (2017-05-06)
Macromolecule-antitumour drug conjugates can reach tumour sites specifically via the enhanced permeability and retention (EPR) effect. It is desirable to release the drug efficiently from the conjugate at acidic pH in the tumour tissue or in the endosomes of cancer
Jae Kyoo Lee et al.
Quarterly reviews of biophysics, 48(4), 437-444 (2015-11-06)
Using high-resolution mass spectrometry, we have studied the synthesis of isoquinoline in a charged electrospray droplet and the complexation between cytochrome c and maltose in a fused droplet to investigate the feasibility of droplets to drive reactions (both covalent and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service