Skip to Content
Merck
All Photos(1)

Key Documents

933708

Sigma-Aldrich

Lithium difluorophosphate

≥99% trace metals basis, acid ≤ 200 ppm, battery grade

Synonym(s):

LiDFP, LiF2PO2, Lithium difluorophosphinate, Lithium phosphorodifluoridate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
LiPO2F2
CAS Number:
Molecular Weight:
107.91
MDL number:
UNSPSC Code:
12161900

grade

battery grade

Quality Level

description

acid ≤ 200 ppm

Assay

≥99% trace metals basis

form

powder

impurities

≤200 ppm acid

cation traces

Ca: ≤5 ppm
Cr: ≤10 ppm
Fe: ≤5 ppm
K: ≤5 ppm
Na: ≤5 ppm
Pb: ≤5 ppm

application(s)

battery manufacturing

General description

Lithium difluorophosphate (LiDFP) is a high-performance additive for lithium-ion batteries. LiDFP is a white powder that is soluble in acetone and dimethoxyethane and slightly soluble (0.4g/100g solvent) in both diethyl carbonate and 1:1 v/v mixtures of dimethylcarbonate and propylene carbonate. Our LiDFP is designed with high-purity and low trace impurities of moisture and acid

Application

Lithium difluorophosphate is a multi-functional additive for high voltage lithium-ion batteries and is often added to lithium hexafluorophosphate (LiPF6)-based electrolytes in carbonate solvents. For example, a recent study showed that a 1 wt% addition of LiDFP to the LiPF6-carbonate electrolyte in a NMC-graphite cell significantly increased capacity retention (97%) after 200 cycles, compared to the same cells without the additive (only 77% capacity retention.)[1] LiDFP has been shown to perform well with nickel-rich cathode materials[2] because it generates a more uniform, extremely stable, and lower resistance solid-electrolyte interphase (SEI) on the cathode surface. This passivation film not only suppresses the subsequent decomposition of the electrolyte, but also prevents the dissolution of transition metal ions from NMC particles, resulting in improved cyclic stability and discharge capacity. In addition, research suggests that LiDPF additive also helps to form a stable and ionically conductive SEI passivation film on lithium metal anodes.[3] Our LiDFP, with high-purity and low moisture and acid impurities, is suitable for research on increasing the energy density and longer cycling performance in lithium-ion batteries and lithium-metal batteries.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium difluorophosphate as a multi-functional electrolyte additive for 4.4V LiNi0.5Co0.2Mn0.3O2/graphite lithium ion batteries
Lei, Qiufen, et al.
Journal of Electroanalytical Chemistry, 846, 113141-113141 (2019)
?Additive engineering for robust interphases to stabilize high-Ni layered structures at ultra-high voltage of 4.8?V
Tan, S., Shadike, Z., Li, J.?et al.
Nature Energy, 7, 484?494-484?494 (2022)
Lithium Difluorophosphate-Based Dual-Salt Low Concentration Electrolytes for Lithium Metal Batteries
Zheng, H., et al.
Advanced Energy Materials, 10, 2001440- 2001440 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service