Skip to Content
Merck
All Photos(1)

Documents

926140

Sigma-Aldrich

(S,S)-Ms-DENEB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H29ClN2O3RuS
CAS Number:
Molecular Weight:
574.10
UNSPSC Code:
12352116
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

reagent type: ligand

functional group

phosphine

Application

(S,S)-Ms-DENEB is an oxo-tethered ruthenium complex for use as an efficient asymmetric hydrogenation catalyst.

Legal Information

DENEB is a trademark of Takasago Intl. Corp.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service