923559
Silver nano ink
aerosol printable
Synonym(s):
Ag NP-based ink, Silver (Ag) Nanoparticle Ink, Silver nano ink
About This Item
Quality Level
form
liquid
contains
50 wt. % Silver content
sheet resistance
1.8 mΩ/sq/25 μm after curing at 150 Celsius for 1h on glass
1.2 mΩ/sq/25 μm after curing at 200 Celsius for 1h on glass
3 mΩ/sq/25 μm after curing at 100 Celsius for 1h on glass
2.3 mΩ/sq/25 μm after curing at 120 Celsius for 1h on glass
color
brownish
viscosity
500-800 mPa.s(20 °C) (@100/s)
solubility
soluble (Alcohol / Glycol)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
to produce antennas for IoT applications (HF, UHF).
This ink has been developed to address the applications which require very low curing temperatures.
Compatible Printing Method: Aerosol jet Optomec
Features and Benefits
- Very high electrical conductivity
- Very low curing temperatures for printing on flexible substrates
- Non CMR ink
- Unchanged conductivity on bending
- Compatible with various curing processes including photonic, IR, low vacuum oven and thermal
- Good adhesion on various flexible substrates (PC, PET, Kapton®, etc)
Usage tips:
- This Conductive Ink needs to be agitated before use
- The ink must be warmed at room temperature before use.
- Dilution with Ethanol can be necessary depending on your printing equipment configuration.
- Clean-up solution: Cleaning solution C-FS21511
Aerosol Jet can print conformal interconnects on 3D surfaces eliminating the need for wire bonding – for example printing electrical connections on 3D stacked die or for LED chip fabrication. Components can also be printed onto 3 dimensional surfaces eliminating the need for a separate substrate thereby reducing the size, thickness and weight of the end product. For example, Aerosol Jet is used to print antennas and sensors that conform to the shape of the underlying substrate such as a cell phone case. The Aerosol Jet process also supports printing on a wide variety of substrates including plastics, ceramics and metallic structures.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service