901311
Polyester bis-MPA dendron 2 NHBoc, 1 azide (core)
generation 1
Synonym(s):
Bis-MPA dendron, PFd-G1-Azide-NH-BOC
About This Item
Recommended Products
description
2 NHBoc, 1 azide (core)
Quality Level
form
solid
mol wt
generation 1
no. Surface Groups
2
color
white to off-white
functional group
azide (core)
storage temp.
−20°C
General description
- Polymer architecture: Dendron
- End Group Functionality: Boc-protected amine
- Boc: tert-Butyloxycarbonyl
- Bis-MPA : 2,2-Bis(hydroxymethyl)propionic acid
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Dendritic molecules' biomedical applications, particularly bis-MPA dendritic scaffolds, hold promise for drug delivery.
Dendritic molecules' biomedical applications, particularly bis-MPA dendritic scaffolds, hold promise for drug delivery.
Dendritic molecules' biomedical applications, particularly bis-MPA dendritic scaffolds, hold promise for drug delivery.
Dendritic molecules' biomedical applications, particularly bis-MPA dendritic scaffolds, hold promise for drug delivery.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service