Skip to Content
Merck
All Photos(1)

Key Documents

82320

Sigma-Aldrich

(±)-Propylene oxide

puriss. p.a., ≥99.5% (GC)

Synonym(s):

(±)-Methyloxirane, 1,2-Epoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O
CAS Number:
Molecular Weight:
58.08
Beilstein:
79763
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

2 (vs air)

Quality Level

vapor pressure

29.43 psi ( 55 °C)
8.59 psi ( 20 °C)

grade

puriss. p.a.

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

1378 °F

expl. lim.

37 %

impurities

≤0.1% water

evapn. residue

≤0.005%

refractive index

n20/D 1.366 (lit.)
n20/D 1.366

bp

34 °C (lit.)

mp

−112 °C (lit.)

density

0.83 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

shipped in

wet ice

storage temp.

2-8°C

SMILES string

CC1CO1

InChI

1S/C3H6O/c1-3-2-4-3/h3H,2H2,1H3

InChI key

GOOHAUXETOMSMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(±)-Propylene oxide (PPO) belongs to the class of monomers known as epoxides or oxiranes. It is used as a raw material in the preparation n of polyurethane foam and polyether polyols, which are used in the production of polyurethane elastomers, polyurethane plastics, adhesives, sealants, and coatings. These materials find applications in industries such as automotive, construction, electronics, and textiles. PPO-based polymers possess flexibility and elasticity as inherent properties, making them well-suited for applications that demand materials with excellent resilience, stretchability, and impact resistance. Additionally, PPO-derived polymers are biocompatible, which makes them suitable for applications in various fields such as biomedical engineering, drug delivery, and tissue engineering.

Application

Propylene oxide can be used as a starting material to synthesize:
  • Poly(3-hydroxybutyrate), a biodegradable and biocompatible polyester, by carbonylative polymerization.
  • Poly (propylene-ram-ε-caprolactone carbonate) (PPCL) and poly (propylene carbonate) (PPC), which are applicable as drug carriers.
Propylene oxide may be used in the production of polyether polyols, propylene glycols, propylene glycol ethers.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-36.4 °F - Equilibrium method

Flash Point(C)

-38 °C - Equilibrium method

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Josep Casamada Ribot et al.
Chemical communications (Cambridge, England), 46(37), 6971-6973 (2010-08-24)
The aqueous gelation of a quaternary ammonium oligo(propylene oxide)-based ionic liquid yields ion gels with a reverse thermoresponsive behavior (i.e., mechanical moduli and viscosity increase with temperature) and enhanced ionic conductivities.
Javix Thomas et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(16), 4582-4587 (2011-03-25)
Chirality recognition in the hydrogen-bonded glycidol···propylene oxide complex has been studied by using rotational spectroscopy and ab initio calculations. An extensive conformational search has been performed for this binary adduct at the MP2/6-311++G(d,p) level of theory and a total of
Sascha Brunke et al.
PLoS pathogens, 10(10), e1004478-e1004478 (2014-10-31)
Candida glabrata is one of the most common causes of candidemia, a life-threatening, systemic fungal infection, and is surpassed in frequency only by Candida albicans. Major factors contributing to the success of this opportunistic pathogen include its ability to readily
Aude Bouchet et al.
The Journal of organic chemistry, 76(10), 4178-4181 (2011-04-08)
ECD and NMR experiments show that the complexation of propylene oxide (PrO) within the cavity of an enantiopure water-soluble cryptophane 1 in NaOH solution is enantioselective and that the (R)-PrO@PP-1 diastereomer is more stable than the (S)-PrO@PP-1 diastereomer with a
Christian M Hammer et al.
Journal of cataract and refractive surgery, 41(6), 1279-1288 (2015-07-21)
To assess the suitability of a new 345 nm ultraviolet (UV) femtosecond laser for refractive surgery. Department of Ophthalmology, University of Erlangen-Nürnberg, Erlangen, Germany. Experimental study. Twenty-five porcine corneas were used for stromal flap or lamellar bed creation (stromal depth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service