Skip to Content
Merck
All Photos(1)

Key Documents

774375

Sigma-Aldrich

2-[(7-Bromo-2,1,3-benzothiadiazol-4-yl)methylene]malononitrile

≥97%

Synonym(s):

2-[(7-Bromo-2,1,3-benzothiadiazol-4-yl)methylene]propanedinitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H3BrN4S
Molecular Weight:
291.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

solid

mp

184-188 °C

storage temp.

2-8°C

SMILES string

Brc1ccc(\C=C(/C#N)C#N)c2nsnc12

InChI

1S/C10H3BrN4S/c11-8-2-1-7(3-6(4-12)5-13)9-10(8)15-16-14-9/h1-3H

InChI key

ZNWYPDKJWGLSQU-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service