Skip to Content
Merck
All Photos(1)

Key Documents

77102

Sigma-Aldrich

4-Pentylphenol

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16O
CAS Number:
Molecular Weight:
164.24
Beilstein:
1364424
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

form

solid

density

0.960 g/mL at 20 °C (lit.)

SMILES string

CCCCCc1ccc(O)cc1

InChI

1S/C11H16O/c1-2-3-4-5-10-6-8-11(12)9-7-10/h6-9,12H,2-5H2,1H3

InChI key

ZNPSUQQXTRRSBM-UHFFFAOYSA-N

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Isobe et al.
Archives of toxicology, 91(11), 3543-3550 (2017-05-14)
4-tert-Octylphenol (4-tOP) is an endocrine-disrupting chemical. It is mainly metabolized into glucuronide by UDP-glucuronosyltransferase (UGT) enzymes in humans. The purpose of this study was to assess inter-individual variability in and the possible roles of UGT isoforms in hepatic 4-tOP glucuronidation
Marta Pastor-Belda et al.
Food chemistry, 221, 76-81 (2016-12-17)
Magnetic solid phase extraction (MSPE) with cobalt ferrite nanoparticles coated with oleic acid is described for the determination of alkylphenols (APs), 4-tert-butylphenol (TBP), 4-pentylphenol (PP), 4-hexylphenol (HP), 4-tert-octylphenol (TOP), 4-n-octylphenol (OP) and 4-nonylphenol (NP) in baby foods using gas chromatography

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service