Skip to Content
Merck
All Photos(1)

Key Documents

764825

Sigma-Aldrich

Poly(ethylene glycol) methyl ether-block-poly(lactide-co-glycolide)

PEG Mn 2,000, PLGA Mn 4,500

Synonym(s):

PEG-PLGA, Polyethylene glycol, mPEG-b-PLGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H[(C3H4O2)x(C2H2O2)y]mO[C2H4O]nCH3
UNSPSC Code:
12162002
NACRES:
NA.23

form

pellets

feed ratio

lactide:glycolide 65:35

mol wt

PEG Mn 2,000
PLGA Mn 4,500
average Mn 6,500 (total)

degradation timeframe

1-4 weeks

transition temp

Tm 241-246 °C

PDI

≤2.0

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Amphiphilic block copolymers (AmBC) are made up of two chemically different homopolymer blocks. One of the block is hydrophilic and the other one is hydrophobic. These macromolecules have the properties to self-assemble when dissolved in an aqueous media. PEG-PLGA is one the most commonly used biodegradable amphiphilic block copolymers for drug delivery applications. PEG is the hydrophilic part and PLGA is the hydrophobic part.

Application

Used in the synthesis of targeted nanoparticles which are used for differential delivery and controlled release of drugs.
forming "stealth" pegylated microparticles

Features and Benefits

  • Good biocompatibility, low immunogenicity and good degradability.
  • Properties can be easily modulated by changing the block copolymer segment sizes to suit a particular application.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermosensitive self-assembling block copolymers as drug delivery systems.
Bonacucina, G., Cespi, M., Mencarelli, G., Giorgioni, G., & Palmieri, G. F.
Polymer, 3(2), 779-811 (2011)
PLGA-PEG Encapsulated sitamaquine nanoparticles drug delivery system against Leishmania donovani
Kumara, R., Sahoo, G. C., Pandeya, K., Dasa, V. N. R., Yousuf, M., Ansaria, S. R., & Dasa, P.
Journal of Scientific and Innovative Research, 3(1), 85-90 (2014)
Frank Gu et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(7), 2586-2591 (2008-02-15)
There has been progressively heightened interest in the development of targeted nanoparticles (NPs) for differential delivery and controlled release of drugs. Despite nearly three decades of research, approaches to reproducibly formulate targeted NPs with the optimal biophysicochemical properties have remained

Articles

Micelle formation addresses low solubility in IV drug delivery, overcoming clinical limitations.

Micelle formation addresses low solubility in IV drug delivery, overcoming clinical limitations.

Micelle formation addresses low solubility in IV drug delivery, overcoming clinical limitations.

Micelle formation addresses low solubility in IV drug delivery, overcoming clinical limitations.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service