753122
Calcium(II) bis(trifluoromethanesulfonimide)
95%
Synonym(s):
Ca(NTf2)2, Calcium triflimide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C4CaF12N2O8S4
CAS Number:
Molecular Weight:
600.37
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
form
powder
mp
>300 °C
SMILES string
FC(F)(F)S(=O)(=O)N([Ca]N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F
InChI
1S/2C2F6NO4S2.Ca/c2*3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;/q2*-1;+2
InChI key
WFABOCFDABTAPE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Calcium(II) bis(trifluoromethanesulfonimide) also known as Calcium triflimide (Ca(NTf2)2), is widely employed as a Lewis acid catalyst in organic synthesis. It facilitates a range of transformations, including Friedel-Crafts alkylation and sulfonamide synthesize.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Calcium-catalyzed Friedel--Crafts addition of 1-methylindole to activated cyclopropanes
Dulin CC, et al.
Tetrahedron Letters, 55(38), 5280-5282 (2014)
Friedel-Crafts alkylation of benzo [b] furan with activated cyclopropanes catalyzed by a calcium (II) complex
Tetrahedron Letters, 59(1), 18-21 (2018)
Sulfonamide synthesis via calcium triflimide activation of sulfonyl fluorides
Organic Letters, 20(13), 3943-3947 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service