Skip to Content
Merck
All Photos(1)

Key Documents

702714

Sigma-Aldrich

Lithium borohydride solution

0.5 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiBH4
CAS Number:
Molecular Weight:
21.78
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reagent type: reductant

concentration

0.5 M in diethyl ether

density

0.719 g/mL at 25 °C

SMILES string

[Li+].[H][B-]([H])([H])[H]

InChI

1S/BH4.Li/h1H4;/q-1;+1

InChI key

UUKMSDRCXNLYOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lithium borohydride is a commonly used reducing agent. It can also be used a reactant for:
  • Preparation of gallium, indium, rhenium and zinc tris(mercaptoimidazolyl)hydroborato complexes.
  • Mechano-chemical metathesis reactions.
  • Noncatalytic hydrolysis for hydrogen generation.
  • Growth of large gold monolayer protected-clusters.
  • Anion substitution reactions.
  • Dehydrogenation reactions.

Reactant for:
  • Preparation of gallium, indium, rhenium and zinc tris(mercaptoimidazolyl)hydroborato complexes
  • Mechano-chemical metathesis reactions
  • Noncatalytic hydrolysis for hydrogen generation
  • Growth of large gold monolayer protected-clusters
  • Anion substitution reactions
  • Dehydrogenation reactions

Other Notes

Material may form precipitate on standing, which does not affect its use.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Controlled growth and catalytic activity of gold monolayer protected clusters in presence of borohydride salts.
Dasog M, et al.
Chemical Communications (Cambridge, England), 47(30), 8569-8571 (2011)
Effect of hydrogen back pressure on dehydrogenation behavior of LiBH4-based reactive hydride composites.
Shim J H, et al.
The Journal of Physical Chemistry Letters, 1(1), 59-63 (2009)
Mechano-chemical reactions in LiBH4+ VCln (n= 2 and 3) mixtures.
Llamas-Jansa I, et al.
J. Alloy Compounds, 509, S684-S687 (2011)
Hydrogen generation from noncatalytic hydrolysis of LiBH4/NH3BH3 mixture for fuel cell applications.
Weng B, et al.
International Journal of Hydrogen Energy, 36(17), 10870-10876 (2011)
Synthesis and structural characterization of tris (2-mercapto-1-adamantylimidazolyl) hydroborato complexes: A sterically demanding tripodal [S3] donor ligand.
Yurkerwich K, et al.
Inorganic Chemistry, 50(24), 12284-12295 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service