Skip to Content
Merck
All Photos(1)

Key Documents

686581

Sigma-Aldrich

Hyperbranched bis-MPA polyester-32-hydroxyl, generation 3

≥97%

Synonym(s):

ALH-32-OH, Hyperbranched Polymer, Hyperpolymer, PFH-32-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C155H256O93
CAS Number:
Molecular Weight:
3607.64
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

hydroxyl surface groups

Assay

≥97%

form

powder (lyophilized)

mol wt

generation 3

no. Surface Groups

32 (avg.)

storage temp.

2-8°C

SMILES string

CC(CO)(CO)C(=O)OCC(C)(COC(=O)C(C)(CO)CO)C(=O)OCC(C)(COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)C(=O)OCCOCCOCC(COCCOC(=O)C(C)(COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)(COCCOC(=O)C(C)(COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COCCOC(=O)C(C)(COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO)COC(=O)C(C)(COC(=O)C(C)(CO)CO)COC(=O)C(C)(CO)CO

InChI

1S/C155H256O93/c1-127(39-156,40-157)99(188)225-79-147(21,80-226-100(189)128(2,41-158)42-159)119(208)241-71-143(17,72-242-120(209)148(22,81-227-101(190)129(3,43-160)44-161)82-228-102(191)130(4,45-162)46-163)115(204)221-35-31-216-29-30-217-95-155(96-218-32-36-222-116(205)144(18,73-243-121(210)149(23,83-229-103(192)131(5,47-164)48-165)84-230-104(193)132(6,49-166)50-167)74-244-122(211)150(24,85-231-105(194)133(7,51-168)52-169)86-232-106(195)134(8,53-170)54-171,97-219-33-37-223-117(206)145(19,75-245-123(212)151(25,87-233-107(196)135(9,55-172)56-173)88-234-108(197)136(10,57-174)58-175)76-246-124(213)152(26,89-235-109(198)137(11,59-176)60-177)90-236-110(199)138(12,61-178)62-179)98-220-34-38-224-118(207)146(20,77-247-125(214)153(27,91-237-111(200)139(13,63-180)64-181)92-238-112(201)140(14,65-182)66-183)78-248-126(215)154(28,93-239-113(202)141(15,67-184)68-185)94-240-114(203)142(16,69-186)70-187/h156-187H,29-98H2,1-28H3

InChI key

IEXLGBFSIFJFKP-UHFFFAOYSA-N

General description

calculated mol. wt. 3,608

Other Notes

Bis-MPA =2,2-Bis(hydroxyl-methyl)propionic acid

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldobenedetto Zotti et al.
Nanomaterials (Basel, Switzerland), 10(2) (2020-01-26)
The effects of the addition of an aromatic hyperbranched polyester (AHBP) on thermal, mechanical, and fracture toughness properties of a thermosetting resin system were investigated. AHBP filler, synthesized by using a bulk poly-condensation reaction, reveals a glassy state at room

Articles

Dendrons in nanomaterials - Find features and benefits of polyester dendrons and hyperbranched polymer products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service