Skip to Content
Merck
All Photos(1)

Documents

674648

Sigma-Aldrich

(S)-(−)-2-Methyl-CBS-oxazaborolidine solution

1 M in THF

Synonym(s):

α,α-Diphenyl-L-prolinol methylboronic acid cycl-amide ester, (S)-1-Methyl-3,3-diphenyl-tetrahydro-pyrrolo[1,2c][1,3,2]oxazaborole, (S)-3,3-Diphenyl-1-methyltetrahydro-3H-pyrrolo[1,2-c][1,3,2]oxazaborole, (S)-Tetrahydro-1-methyl-3,3-diphenyl-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20BNO
CAS Number:
Molecular Weight:
277.17
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

concentration

1 M in THF

refractive index

n20/D 1.460

density

0.942 g/mL at 25 °C

SMILES string

CB1OC([C@@H]2CCCN12)(c3ccccc3)c4ccccc4

InChI

1S/C18H20BNO/c1-19-20-14-8-13-17(20)18(21-19,15-9-4-2-5-10-15)16-11-6-3-7-12-16/h2-7,9-12,17H,8,13-14H2,1H3/t17-/m0/s1

InChI key

VMKAFJQFKBASMU-KRWDZBQOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The CBS (Corey-Bakshi-Shibata) oxazaborolidine catalyst has been used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2-symmetrical ferrocenyl diols, and propargyl alcohols.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

5.0 °F - closed cup

Flash Point(C)

-15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Singh, V. K.
Synthesis, 605-605 (1992)
Asymmetric Reduction. A Convenient Method for the Reduction of Alkynyl Ketones.
Kathlyn A. Parker et al.
The Journal of organic chemistry, 61(9), 3214-3217 (1996-05-03)
E.J. Corey et al.
Tetrahedron Letters, 31, 611-611 (1990)
Corey, E. J. et al.
Journal of the American Chemical Society, 109, 5551-5551 (1987)
Cliff R Baar et al.
Journal of the American Chemical Society, 126(26), 8216-8231 (2004-07-01)
A series of enantiopure C1-symmetric metallocenes, [(SiMe2)2[eta5-C5H(CHMe2)2][eta5-C5H2((S)-CHMeCMe3)]]ZrCl2, (S)-2, [(SiMe2)2[eta5-C5H(CHEt2)2][eta5-C5H2((S)-CHMeCMe3)]]ZrCl2, (S)-6, and [(SiMe2)2[eta5-C5HCy2][eta5-C5H2((S)-CHMeCMe3)]]ZrCl2, (S)-7 (Cy = cyclohexyl), zirconocene dichlorides that have an enantiopure methylneopentyl substituent on the "upper" cyclopentadienyl ligand, and diastereomerically pure precatalysts, [(SiMe2)2[eta5-C5H((S)-CHMeCy)(CHMe2)][eta5-C5H3]]ZrCl2, (S)-8a and (S)-8b, which have an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service