Skip to Content
Merck
All Photos(1)

Key Documents

662747

Sigma-Aldrich

3-Bromo-2-butanone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7BrO
CAS Number:
Molecular Weight:
151.00
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.460

density

1.416 g/mL at 25 °C

SMILES string

CC(Br)C(C)=O

InChI

1S/C4H7BrO/c1-3(5)4(2)6/h3H,1-2H3

InChI key

BNBOUFHCTIFWHN-UHFFFAOYSA-N

Application

3-Bromo-2-butanone can be used as a reactant to synthesize:
  • N



  • -(4,5-Dimethyl-1H-imidazol-2-yl)acetamide by reacting with N-acetylguanidine under acidic conditions.
  • Oxazole derivatives by silver triflate catalyzed cyclization reaction with various amides.
  • 2-bromo-5,6-dimethyl[1,3]oxazolo[3,2-b][1,2,4]triazole by treating with 3,5-dibromo-1,2,4-triazole in the presence DBU.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

123.8 °F

Flash Point(C)

51 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[1, 3] Oxazolo [3, 2-b][1, 2, 4] triazoles: a versatile synthesis of a novel heterocycle
Ball Catherine, et al.
Tetrahedron Letters, 51(30), 3907-3909 (2010)
Synthesis of 2, 4-and 2, 4, 5-substituted oxazoles via a silver triflate mediated cyclization
Bailey JL and Sudini RR
Tetrahedron Letters, 55(27) (2014)
Chai Hoon Soh et al.
Journal of combinatorial chemistry, 10(1), 118-122 (2007-12-28)
A microwave-assisted protocol was developed for the construction of di- and monosubstituted 2-aminoimidazoles. The two-step reaction involves the synthesis of N-(1H-imidazol-2-yl)acetamides from readily available alpha-haloketones and N-acetylguanidine, followed by deacetylation. Significant rate enhancement was observed for both steps of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service