Skip to Content
Merck
All Photos(2)

Key Documents

647640

Sigma-Aldrich

Zirconium(IV) chloride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Tetrachlorozirconium, Zirconium tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZrCl4
CAS Number:
Molecular Weight:
233.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

vapor pressure

1 mmHg ( 190 °C)

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: zirconium

impurities

<50 ppm hafnium

transition temp

sublimation point 331 °C

density

2.8 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Cl[Zr](Cl)(Cl)Cl

InChI

1S/4ClH.Zr/h4*1H;/q;;;;+4/p-4

InChI key

DUNKXUFBGCUVQW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Zirconium(IV) chloride (ZrCl4) is a Lewis acid catalyst, which has low toxicity. It is a moisture resistant material that is used as a catalyst in organic transformations.

Application

ZrCl4 can be used as a catalyst for a variety of organic syntheses, such as Friedel-Crafts reaction, condensation reaction and other reduction reactions.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik A Wu et al.
Nature communications, 12(1), 1256-1256 (2021-02-25)
Rechargeable solid-state sodium-ion batteries (SSSBs) hold great promise for safer and more energy-dense energy storage. However, the poor electrochemical stability between current sulfide-based solid electrolytes and high-voltage oxide cathodes has limited their long-term cycling performance and practicality. Here, we report
Synthesis of 2, 3-dihydroquinazolin-4 (1H)-ones catalyzed by zirconium (IV) chloride as a mild and efficient catalyst
Abdollahi-Alibeik M and Shabani E
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 22(10), 1163-1166 (2011)
Applications of zirconium (IV) chloride in organic synthesis
Smitha G, et al.
Synthesis, 2008(06), 829-855 (2008)
Ilanit Doron-Mor et al.
Langmuir : the ACS journal of surfaces and colloids, 20(24), 10727-10733 (2004-11-17)
Coordination self-assembly of bishydroxamate-based metal-organic multilayers on gold employing a layer-by-layer (LbL) approach was investigated. It is shown that the solution chemistry of the participating metal ion has a marked influence on the composition and properties of the multilayers. Use
Giles W Theaker et al.
Dalton transactions (Cambridge, England : 2003), (48)(48), 6883-6885 (2008-12-04)
A group of readily available zirconium catalysts incapable of ethene-co-styrene polymerization are remarkably active and selective for the production of the new polymer ethene-co-tert-butylstyrene via a single site mechanism.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service