Skip to Content
Merck
All Photos(1)

Key Documents

62141

Sigma-Aldrich

Linalool oxide

mixture of isomers, ≥97.0% (GC)

Synonym(s):

2-(5-Methyl-5-vinyltetrahydro-1-furyl)-2-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
Beilstein:
117527
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

density

0.945 g/mL at 20 °C (lit.)

SMILES string

CC(C)(O)C1CCC(C)(O1)C=C

InChI

1S/C10H18O2/c1-5-10(4)7-6-8(12-10)9(2,3)11/h5,8,11H,1,6-7H2,2-4H3

InChI key

BRHDDEIRQPDPMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Linalool oxide is a monoterpenoid compound, commonly found in some species of the aromatic plants. It can be obtained from linalool either by oxidation or via biotransfomation using the fungus Aspergillus niger. It is always present as a mixture of both cis and trans forms.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Anxiolytic-like effects of inhaled linalool oxide in experimental mouse anxiety models"
Souto-Maior NF, et al.
Pharmacology, Biochemistry, and Behavior, 100(02), 259-263 (2011)
Apsimon J
Total Synthesis of Natural Products, 2 (2009)
Bao-Ting Yu et al.
Parasites & vectors, 8, 598-598 (2015-11-19)
Most mosquito species need to obtain sugar from host plants. Little is known about the chemical cues that Culex pipiens pallens use during their orientation to nectar host plants. In this study, we investigated the behavioural responses of female Cx.
Paroma Mitra et al.
Pest management science, 77(1), 285-299 (2020-07-23)
The viviparous aphid Aphis craccivora Koch (Hemiptera: Aphididae) is a serious threat to the crop yield of Lathyrus sativus L. (Fabaceae), commonly known as grass pea. The synthetic insecticides applied to control this insect pest are not safe for the
Vincent O Nyasembe et al.
Parasites & vectors, 8, 581-581 (2015-11-11)
Lack of effective vaccines and therapeutics for important arboviral diseases such as Rift Valley fever (RVF) and dengue, necessitates continuous monitoring of vector populations for infections in them. Plant-based lures as surveillance tools has the potential of targeting mosquitoes of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service