Skip to Content
Merck
All Photos(1)

Documents

600113

Sigma-Aldrich

L-Arginine-15N4 hydrochloride

98 atom % 15N, 97% (CP)

Synonym(s):

SILAC Amino Acid, (S)-(+)-Arginine hydrochloride-15N4, 15N Labeled arginine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H215NC(15NH)15NH(CH2)3C(15NH2)CO2H · HCl
CAS Number:
Molecular Weight:
214.64
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 15N

Assay

≥97% (Chiral Purity)
97% (CP)

form

solid

optical activity

[α]20/D +22°, c = 12 in 10% HCl

technique(s)

bio NMR: suitable
protein expression: suitable

mp

226-230 °C (dec.) (lit.)

mass shift

M+4

SMILES string

Cl[H].[15NH2][C@@H](CCC[15NH]C([15NH2])=[15NH])C(O)=O

InChI

1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H/t4-;/m0./s1/i7+1,8+1,9+1,10+1;

InChI key

KWTQSFXGGICVPE-JYJSWXFRSA-N

Related Categories

General description

Arginine is an amino acid with a basic side group. In liver, arginine is cleaved to ornithine and urea. Isotope labeled amino acids are required for the production of isotopically labeled proteins.

Application

L-Arginine-15N4 hydrochloride has been used as a supplement for growing rat hybridoma to generate 15N-labeled monoclonal antibody against GPIHBP1 (glycosylphosphatidylinositol-anchored HDL binding protein 1).

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harvey RA, et al.
Biochemistry (Lippincott's illustrated reviews) (2011)
High-resolution imaging of dietary lipids in cells and tissues by NanoSIMS analysis.
Jiang H, et al.
Journal of Lipid Research, 55, 2156-2166 (2014)
Isotope labeling for solution and solid-state NMR spectroscopy of membrane proteins.
Verardi R, et al.
Advances in Experimental Medicine and Biology, 992, 35-62 (2012)
Incorporation of radioactive amino acids into proteins of the microsome fraction of guinea-pig liver in a cell-free system.
SIMKIN JL and WORK TS
The Biochemical Journal, 67, 617-624 (1957)
The incorporation of radioactive amino acids into proteins of the microsome fraction of guinea-pig liver at very short time intervals after administration.
Simkin JL and Sutton CR
The Biochemical Journal, 77, 489-492 (1960)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service