Skip to Content
Merck
All Photos(1)

Documents

594687

Sigma-Aldrich

α-Sexithiophene

Synonym(s):

6T, Sexithiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H14S6
CAS Number:
Molecular Weight:
494.76
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

mp

290 °C (dec.) (lit.)

Orbital energy

HOMO 5.3 eV (opt)
LUMO 3.1 eV (opt)

OPV Device Performance

ITO/6T/C60/BCP/Mg:Ag

  • Short-circuit current density (Jsc): 0.8 mA/cm2
  • Open-circuit voltage (Voc): 0.4 V
  • Fill Factor (FF): 0.46
  • Power Conversion Efficiency (PCE): 0.52 %

ITO/6T/C70/BCP/Mg:Ag
  • Short-circuit current density (Jsc): 4.1 mA/cm2
  • Open-circuit voltage (Voc): 0.41 V
  • Fill Factor (FF): 0.55
  • Power Conversion Efficiency (PCE): 0.92 %

ITO/6T:C70 (1:5)/BCP/Mg:Ag
  • Short-circuit current density (Jsc): 9.2 mA/cm2
  • Open-circuit voltage (Voc): 0.58 V
  • Fill Factor (FF): 0.45
  • Power Conversion Efficiency (PCE): 2.38 %

semiconductor properties

P-type (mobility=0.075 cm2/V·s)

SMILES string

c1csc(c1)-c2ccc(s2)-c3ccc(s3)-c4ccc(s4)-c5ccc(s5)-c6cccs6

InChI

1S/C24H14S6/c1-3-15(25-13-1)17-5-7-19(27-17)21-9-11-23(29-21)24-12-10-22(30-24)20-8-6-18(28-20)16-4-2-14-26-16/h1-14H

InChI key

KUJYDIFFRDAYDH-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service