Skip to Content
Merck
All Photos(2)

Documents

553123

Sigma-Aldrich

Tetrakis(ethylmethylamido)hafnium(IV)

≥99.99% trace metals basis

Synonym(s):

TEMAH, Tetrakis(ethylmethylamino)hafnium(IV)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)(C2H5)N]4Hf
CAS Number:
Molecular Weight:
410.90
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99% trace metals basis

form

liquid

reaction suitability

core: hafnium

impurities

Purity excludes ~2000 ppm Zirconium

bp

78 °C/0.01 mmHg (lit.)

mp

<-50 °C

density

1.324 g/mL at 25 °C (lit.)

SMILES string

CCN(C)[Hf](N(C)CC)(N(C)CC)N(C)CC

InChI

1S/4C3H8N.Hf/c4*1-3-4-2;/h4*3H2,1-2H3;/q4*-1;+4

InChI key

NPEOKFBCHNGLJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrakis(ethylmethylamido)hafnium(IV) (TEMAH) is a colorlessliquid that is sensitive to water and air. It freezes at -50 °C and boilsaround 78 °C at 0.1 Torr.

Application

TEMAH is used as a precursor for atomic layer deposition (ALD)of hafnium oxide (HfO2) thin films. Because HfO2 has a high dielectric constant of 16-25, it is commonly used as a dielectric film in semiconductor fabrication.

TEMAH is ideal for ALD because of its low boiling point and its reactivity with water and ozone. Most importantly, its adsorption is self-limiting on a number of substrates including glass, indium-tin oxide(ITO), and Si(100). Researchers have also used it to deposit thin films ofHfO2 on 2D materials, like MoS2.


TEMAH is also useful precursor in the synthesis of ferroelectric hafnium zirconium oxide and Hf1-xZrxO2 thin films on MoS2 phototransistors. Researchers have also deposited thin films of hafnium nitride (Hf3N4) by ALD alternatively pulsing TEMAH and ammonia.

Features and Benefits

  • Thermally stable.
  • It has sufficient volatility and is suitable for use in vapor deposition.
  • Completely self-limiting surface reactions.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atomic Layer Deposition of Hafnium Dioxide Films from Hafnium Tetrakis(ethylmethylamide) and Water.
Kukli K, et al.
Chem. Vap. Deposition, 8, 199-204 (2002)
Jaehyun Yang et al.
ACS applied materials & interfaces, 5(11), 4739-4744 (2013-05-21)
We report on the effect of oxygen plasma treatment of two-dimensional multilayer MoS2 crystals on the subsequent growth of Al2O3 and HfO2 films, which were formed by atomic layer deposition (ALD) using trimethylaluminum and tetrakis-(ethylmethylamino)hafnium metal precursors, respectively, with water
Luqi Tu et al.
Nature communications, 11(1), 101-101 (2020-01-05)
Sensitive photodetection is crucial for modern optoelectronic technology. Two-dimensional molybdenum disulfide (MoS2) with unique crystal structure, and extraordinary electrical and optical properties is a promising candidate for ultrasensitive photodetection. Previously reported methods to improve the performance of MoS2 photodetectors have focused
Atomic Layer Deposition of Insulating Hafnium and Zirconium Nitrides.
Becker J S, et al.
Chemistry of Materials, 3497?3501-3497?3501 (2004)

Articles

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service