Skip to Content
Merck
All Photos(2)

Documents

537403

Sigma-Aldrich

2-(Methacryloyloxy)ethyl acetoacetate

95%, contains BHT as stabilizer

Synonym(s):

AAEM, (2-Acetoacetoxy)ethyl methacrylate, Ethylene glycol monoacetoacetate monomethacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CO2CH2CH2O2CC(CH3)=CH2
CAS Number:
Molecular Weight:
214.22
Beilstein:
2099809
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

contains

BHT as stabilizer

refractive index

n20/D 1.456 (lit.)

bp

100 °C/0.8 mmHg (lit.)

density

1.122 g/mL at 25 °C (lit.)

SMILES string

CC(=O)CC(=O)OCCOC(=O)C(C)=C

InChI

1S/C10H14O5/c1-7(2)10(13)15-5-4-14-9(12)6-8(3)11/h1,4-6H2,2-3H3

InChI key

IBDVWXAVKPRHCU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-(Methacryloyloxy)ethyl acetoacetate is a methylene active compound that may be used in the synthesis of 2-(methacryloyloxy)ethyl 6-methyl-2-oxo-4-phenyl-1,2,3,4- tetrahydropyrimidine-5-carboxylate. It may be used as a polymerizable chelating agent for metal alkoxides to form polymerizable oxide precursors.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phat Tran et al.
Biomedicines, 8(3) (2020-03-21)
The dressing material of a wound plays a key role since bacteria can live in the bandage and keep re-infecting the wound, thus a bandage is needed that blocks biofilm in the bandage. Using an in vivo wound biofilm model
Large Area, Facile Oxide Nanofabrication via Step-and-Flash Imprint Lithography of Metal?Organic Hybrid Resins.
Dinachali S S
ACS Applied Materials & Interfaces, 5(24), 13113-13123 (2013)
Juntao Wei et al.
Bioresource technology, 239, 482-489 (2017-05-26)
In this work, the influences of gasification temperature and blended ratio on co-gasification reactivity and synergy of Shenfu bituminous coal (SF) and municipal solid waste-derived hydrochar (HTC) were investigated using TGA. Additionally, active alkaline and alkaline earth metal (AAEM) transformation
Synthesis of some tetrahydropyrimidine-5-carboxylates, determination of their metal chelating effects and inhibition profiles against acetylcholinesterase, butyrylcholinesterase and carbonic anhydrase.
Sujayev A
Journal of Enzyme Inhibition and Medicinal Chemistry, 31(6), 1531-1539 (2016)
J Wang et al.
Nanoscale research letters, 4(3), 240-246 (2008-01-01)
This work demonstrates the synthesis of core-shell ZrO2/PAAEM/PS nanoparticles through a combination of sol-gel method and emulsifier-free emulsion polymerizaiton. By this method, the modified nanometer ZrO2cores were prepared by chemical modification at a molecular level of zirconium propoxide with monomer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service