Skip to Content
Merck
All Photos(1)

Key Documents

486477

Sigma-Aldrich

Chloroacetyl chloride-1-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH213COCl
CAS Number:
Molecular Weight:
113.94
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % 13C

refractive index

n20/D 1.453 (lit.)

bp

105-106 °C (lit.)

mp

−22 °C (lit.)

density

1.430 g/mL at 25 °C

mass shift

M+1

SMILES string

ClC[13C](Cl)=O

InChI

1S/C2H2Cl2O/c3-1-2(4)5/h1H2/i2+1

InChI key

VGCXGMAHQTYDJK-VQEHIDDOSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A - STOT RE 1

Target Organs

Lungs

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service