Skip to Content
Merck
All Photos(2)

Key Documents

481882

Sigma-Aldrich

Cadmium sulfate

≥99.99% trace metals basis

Synonym(s):

Cadmium monosulfate, Cadmium sulfate (Cd(SO4 ))

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSO4
CAS Number:
Molecular Weight:
208.47
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: cadmium

impurities

≤100 ppm Trace Metal Analysis

SMILES string

[Cd++].[O-]S([O-])(=O)=O

InChI

1S/Cd.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

QCUOBSQYDGUHHT-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Applied in the formation of novel two-dimensional Cd-SCN coordination solids with unusual and tailorable, checkerboard- or herringbone-patterned structures these structures are important steps toward technologically useful materials.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Target Organs

Bone,Kidney,Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela Rubio et al.
Nucleic acids research, 49(1), 383-399 (2020-12-15)
Translational control is essential in response to stress. We investigated the translational programmes launched by the fission yeast Schizosaccharomyces pombe upon five environmental stresses. We also explored the contribution of defence pathways to these programmes: The Integrated Stress Response (ISR)
Hong Zhang et al.
Inorganic chemistry, 38(5), 886-892 (2001-10-24)
This paper reports the syntheses, characterization, and structures of two new two-dimensional cadmium-thiocyanate coordination solids [(12C4)(2)Cd][Cd(2)(SCN)(6)] (1) and [(12C4)(2)Cd][Cd(3)(SCN)(8)] (2), where 12C4 denotes 12-crown-4, C(8)H(16)O(4). 1 crystallizes in a monoclinic unit cell, space group C2/c with lattice parameters a =
Shurong Tang et al.
Biosensors & bioelectronics, 42, 608-611 (2012-12-25)
A novel highly sensitive and selective electrochemical sensing system based on DNAzyme and rolling circle amplification (RCA) for the determination of Pb²⁺ was developed in the present study. Firstly, the DNAzyme catalytic strands were immobilized onto magnetic beads surface and
Renguo Xie et al.
Journal of the American Chemical Society, 131(42), 15457-15466 (2009-09-25)
The initial formation of semiconductor nanocrystals/nanoclusters, that is, nucleation in the classic literature, was examined both theoretically and experimentally. An experimental method based on determining the initial reaction rate for the formation of nanocrystals/nanoclusters with fixed size and size distribution
Electric-field enhancement of photovoltaic devices: a third reason for the increase in the efficiency of photovoltaic devices by carbon nanotubes.
Wonjoo Lee et al.
Advanced materials (Deerfield Beach, Fla.), 22(20), 2264-2267 (2010-03-31)

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service