Skip to Content
Merck
All Photos(1)

Documents

464597

Sigma-Aldrich

Ammonium zirconium(IV) carbonate solution

in H2O, contains 1-2% tartaric acid as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

liquid

contains

1-2% tartaric acid as stabilizer

concentration

in H2O
13-22% Zr (gravimetric)

mp

−4 °C

density

1.38 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

N.N.OC(=O)O[Zr](O)(O)OC(O)=O

InChI

1S/2CH2O3.2H3N.2H2O.Zr/c2*2-1(3)4;;;;;/h2*(H2,2,3,4);2*1H3;2*1H2;/q;;;;;;+4/p-4

InChI key

VTZCXKGZGDSBQC-UHFFFAOYSA-J

General description

Ammonium zirconium(IV) carbonate (AZC) solution is an industrial salt based solution that is majorly used in the surface treatment of metals. It may also be used as a cross-linker for water soluble macromolecules.

Application

AZC can be used as a precursor for the preparation of zirconium oxide which can further be used in biomedicine and other nano-based applications. It can be reacted with borax to form an acid-base bifunctional catalyst for selective organic transformation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mesoporous borated zirconia: a solid acid-base bifunctional catalyst
Sinhamahapatra A, et al.
ChemCatChem, 5(1), 331-338 (2013)
Poriferan chitin as a template for hydrothermal zirconia deposition
Wysokowski M, et al.
Frontiers in Marine Science, 7(3) (2013)
Poriferan chitin as a template for hydrothermal zirconia deposition
Wysokowski M, et al.
Frontiers in Marine Science, 7(3), 248-260 (2013)
Extreme biomimetics: formation of zirconium dioxide nanophase using chitinous scaffolds under hydrothermal conditions
Ehrlich H, et al.
Journal of materials chemistry. B, 1(38), 5092-5099 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service