Skip to Content
Merck
All Photos(1)

Key Documents

464287

Sigma-Aldrich

N,N-Dimethyldecylamine

98%

Synonym(s):

1-(Dimethylamino)decane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9N(CH3)2
CAS Number:
Molecular Weight:
185.35
Beilstein:
1738198
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.431 (lit.)

bp

234 °C (lit.)

density

0.778 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCN(C)C

InChI

1S/C12H27N/c1-4-5-6-7-8-9-10-11-12-13(2)3/h4-12H2,1-3H3

InChI key

YWWNNLPSZSEZNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethyldecylamine (DMDA) is a tertiary amine.

Application

N,N-Dimethyldecylamine (DMDA) may be used for the following studies:
  • Pore expansion of the aminopropyl-functionalized ethane-bridged bifunctional periodic mesoporous organosilicas (APEPMOs).
  • Preparation of stationary phase (named QA C10) having a quaternary ammonium embedded between a propyl and a decyl chain.
  • Pore exapansion of monodisperse phenylene-bridged organosilica spheres.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liye Guan et al.
Biotechnology letters, 35(8), 1323-1330 (2013-04-18)
The aminopropyl-functionalized ethane-bridged bifunctional periodic mesoporous organosilicas (APEPMOs) were synthesized by the co-condensation of 1,2-bis (triethoxysilyl) ethane and 3-aminopropyltriethoxysilane in the presence of cationic surfactants octadecyltrimethylammonium chloride in basic medium. The pores of the APEPMOs were expanded with N,N-dimethyldecylamine and
Qiaoxia Liu et al.
Journal of separation science, 35(20), 2685-2692 (2012-09-26)
A stationary phase (named QA C10) with quaternary ammonium embedded between a propyl and a decyl chain was synthesized by immobilization of N,N-dimethyldecylamine on chloropropyl-silica surface. A set of representative neutral, basic, and acidic compounds was employed to evaluate its
Yongping Zhang et al.
Talanta, 81(3), 824-830 (2010-03-20)
Monodisperse phenylene-bridged organosilica spheres show great potential as chromatographic stationary phase. In this paper, the tunable particle size of monodisperse phenylene-bridged organosilica spheres were prepared by co-condensing different proportion of 1,4-bis(triethoxysilyl)benzene (1,4-BTEB) and tetraethylorthosilicate (TEOS), and then pore size was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service