Skip to Content
Merck
All Photos(1)

Key Documents

447005

Sigma-Aldrich

N,N′-Di-tert-butylethylenediamine

98%

Synonym(s):

1,2-Bis(tert-butylamino)ethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CNHCH2CH2NHC(CH3)3
CAS Number:
Molecular Weight:
172.31
Beilstein:
1738432
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.43 (lit.)

bp

196-198 °C (lit.)

density

0.799 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CC(C)(C)NCCNC(C)(C)C

InChI

1S/C10H24N2/c1-9(2,3)11-7-8-12-10(4,5)6/h11-12H,7-8H2,1-6H3

InChI key

KGHYGBGIWLNFAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silylene and Germylene Intermediates in the Reactions of Silole and Germole Dianions with N,N'-Di-tert-butylethylenediimine.
Toulokhonova IS, et al.
European Journal of Inorganic Chemistry, 14, 2344-2349 (2008)
Aluminium amides derived from metallation of N,N'-di-tert-butylethylenediamine.
Gardiner MG, et al.
J. Chem. Soc., Dalton Trans., 22, 4163-4169 (1996)
Meishan Chen et al.
Molecules (Basel, Switzerland), 24(8) (2019-04-21)
In recent years, polyureas with dynamic hindered urea bonds (HUBs), a class of promising biomedical polymers, have attracted wide attention as a result of their controlled hydrolytic properties. The effect of the chemical structures on the properties of polyureas and
Michael G. Gardiner et al.
Inorganic chemistry, 35(13), 4047-4059 (1996-06-19)
The lithiation of N,N'-di-tert-butylethylenediamine by MeLi in benzene has been shown by (1)H NMR spectroscopy to proceed via the partially lithiated species [cis-{Li[&mgr;-N(t-Bu)CH(2)CH(2)N(H)t-Bu]}(2)], 2, and [{Li[N(t-Bu)CH(2)CH(2)N(H)t-Bu]}(2)Li{N(t-Bu)CH(2)CH(2)Nt-Bu}Li], 3, prior to the formation of the dilithiated species {Li[N(t-Bu)CH(2)CH(2)Nt-Bu]Li}, 4. The solid state
New chemistry from the reaction of N, N'-disubstituted ethylenediamines with glyoxal: synthesis of 2-imidazolidinecarboxaldehydes and 1, 4, 6, 9-tetraalkyl-1, 4, 6, 9-tetraaza-5, 10-dioxaperhydroanthracenes.
Willer RL, et al.
The Journal of Organic Chemistry, 50(13), 2368-2372 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service