Skip to Content
Merck
All Photos(1)

Documents

410020

Sigma-Aldrich

Ethylene glycol vinyl ether

97%

Synonym(s):

2-(Vinyloxy)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHOCH2CH2OH
CAS Number:
Molecular Weight:
88.11
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

143 °C (lit.)

density

0.982 g/mL at 25 °C (lit.)

SMILES string

OCCOC=C

InChI

1S/C4H8O2/c1-2-6-4-3-5/h2,5H,1,3-4H2

InChI key

VUIWJRYTWUGOOF-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

E Dinar et al.
Environmental science & technology, 42(3), 793-799 (2008-03-08)
Recent field observations suggest that ammonium salts of organic acids may be very important in accounting for aerosols' properties in many environments. In this study we present laboratory experiments and calculations on the influence of ammonia reaction with organic aerosol
R Z Ikramov et al.
Biulleten' eksperimental'noi biologii i meditsiny, 112(11), 553-555 (1991-11-01)
Application of ultrasound surgical aspirator promotes rapid formation of major cavities with dry inner surface in the depth of hepatic parenchyma. Filling of the latter with sutured copolymer of monovinyl ether of ethyleneglycol in the ratio of one filler volume
Xueliang Yang et al.
Physical chemistry chemical physics : PCCP, 13(48), 21288-21300 (2011-10-26)
The pyrolysis of ethylene glycol vinyl ether (EGVE), an initial product of 1,4-dioxane dissociation, was examined in a diaphragmless shock tube (DFST) using laser schlieren densitometry (LS) at 57 ± 2 and 122 ± 3 Torr over 1200-1800 K. DFST/time-of-flight
Xiaoli Zeng et al.
Carbohydrate polymers, 237, 116139-116139 (2020-04-04)
We first synthesized indomethacin (IND)-grafted dextran copolymer by acetal or ester linkage, which self-assembled with doxorubicin (DOX) into prodrug micelles (IDAC/DOX or IDES/DOX) with the size of ∼200 nm. In vitro drug release test verified IDAC/DOX could trigger more DOX and
Shih-Horng Su et al.
Annals of biomedical engineering, 31(6), 667-677 (2003-06-12)
A bioresorbable, expandable poly(L-lactic acid) stent has been designed, based on a linear, continuous coil array principle, by which multiple furled lobes convert to a single lobe upon balloon expansion, without heating. Stent strength and compliance are sufficient to permit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service