Skip to Content
Merck
All Photos(1)

Key Documents

381667

Sigma-Aldrich

Diethylenetriaminepentaacetic acid gadolinium(III) dihydrogen salt hydrate

97%

Synonym(s):

Gadopentetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20GdN3O10 · xH2O
Molecular Weight:
547.57 (anhydrous basis)
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

core: gadolinium
reagent type: catalyst

mp

129 °C (dec.) (lit.)

SMILES string

[Gd+3].[H]O[H].OC(=O)CN(CCN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C14H23N3O10.Gd.H2O/c18-10(19)5-15(1-3-16(6-11(20)21)7-12(22)23)2-4-17(8-13(24)25)9-14(26)27;;/h1-9H2,(H,18,19)(H,20,21)(H,22,23)(H,24,25)(H,26,27);;1H2/q;+3;/p-3

InChI key

JBUMALASVLVYKI-UHFFFAOYSA-K

Application

Widely used metal ion complex for MRI used in the diagnosis of cerebral tumors, CNS diseases, hepatic tumors, pituitary adenomas, multiple sclerosis, and blood-brain barrier impairment.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Dwyer et al.
Radiology, 163(2), 421-426 (1987-05-01)
To assess the role of magnetic resonance imaging in the evaluation of adrenocorticotropic-hormone (ACTH)-producing pituitary adenomas, and the effect of intravenously administered gadolinium-diethylenetriaminepentaacetic acid (DTPA) on lesion/pituitary contrast, imaging was performed in 13 patients with clinical and chemical evidence of
K Ohtomo et al.
Radiology, 163(1), 27-31 (1987-04-01)
Thirty-six patients with hepatic tumors (28 hepatocellular carcinomas, seven cavernous hemangiomas, one metastatic tumor) were examined with serial magnetic resonance (MR) imaging, after a bolus intravenous injection of 0.05 mmol/kg gadolinium-diethylenetriaminepentaacetic acid. A typical MR imaging pattern for hemangiomas (present
Christof Karmonik et al.
World neurosurgery, 105, 775-782 (2017-06-19)
Hemodynamics in cerebral aneurysms are currently investigated toward clinical efficacy using nonstandardized computational simulation techniques. At the same time, flow patterns and velocities are accessible by 4-dimensional phase contrast magnetic resonance imaging (4D pcMRI). Complexity of protocol design and imaging
Magnetic resonance imaging in neurosarcoidosis.
A Greco et al.
Magnetic resonance imaging, 5(1), 15-21 (1987-01-01)
Evgeniya Goldman et al.
Nanotechnology, 28(43), 43LT01-43LT01 (2017-09-06)
Despite advances in cancer therapy, treating cancer after it has metastasized remains an unmet clinical challenge. In this study we demonstrate that 100 nm liposomes target triple-negative murine breast-cancer metastases post intravenous administration. Metastatic breast cancer was induced in BALB/c

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service