Skip to Content
Merck
All Photos(1)

Documents

361747

Sigma-Aldrich

Chlorobis(trimethylsilyl)methane

97%

Synonym(s):

Bis(trimethylsilyl)chloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3Si]2CHCl
CAS Number:
Molecular Weight:
194.85
Beilstein:
1736681
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.449 (lit.)

bp

57-60 °C/15 mmHg (lit.)

density

0.892 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)C(Cl)[Si](C)(C)C

InChI

1S/C7H19ClSi2/c1-9(2,3)7(8)10(4,5)6/h7H,1-6H3

InChI key

XNJGZHVYPBNLEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chlorobis(trimethylsilyl)methane can be used as a reagent for the preparation of:
  • Para-bis(trimethylsilyl)ethylstyrene (PBTES) monomer, which is used to synthesize corresponding network polymer of styrene.
  • Bis(trimethylsilyl)methyl magnesium chloride (Grignard reagent), which is used in the synthesis of bis(trimethylsilyl) allyl compounds by reacting with alkenyl bromide via the Kumada coupling reaction.
  • N-[Bis(trimethylsilyl)methyl]heterocumulenes.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Synthesis of some silyl mono-and polystyrenes with new properties
Assadi, MG and Hosseinzadeh, F
Designed Monomers and Polymers, 13(2), 181-191 (2010)
David R Williams et al.
Organic letters, 8(20), 4393-4396 (2006-09-22)
Allylation reagents, which possess geminal bis-trimethylsilyl substitution, are readily prepared from E- or Z-alkenyl bromides. The reactivity of 3,3-bis(trimethylsilyl)-2-methyl-1-propene (1) is described and predominantly provides ene reactions with aldehydes to give alcohol 2 in the presence of BF3.OEt2. Alternatively, Sakurai
Synthesis and Reactivity of N-[Bis (trimethylsilyl) methyl] heterocumulenes
Barbaro G, et al.
The Journal of Organic Chemistry, 60(19), 6032-6039 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service