Skip to Content
Merck
All Photos(4)

Documents

307831

Sigma-Aldrich

Sodium chromate

98%

Synonym(s):

Chromium sodium oxide, Disodium chromate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CrO4
CAS Number:
Molecular Weight:
161.97
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

SMILES string

[Na+].[Na+].[O-][Cr]([O-])(=O)=O

InChI

1S/Cr.2Na.4O/q;2*+1;;;2*-1

InChI key

PXLIDIMHPNPGMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Green Synthetic Route of Sodium Chromate: Describes a novel method to produce sodium chromate from ferrochrome, which could lead to more sustainable production practices in the industry (Li et al., 2020).

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xiaomin Ni et al.
Biomolecules, 11(2) (2021-01-28)
Hexavalent chromium [Cr(VI)] pollution is one of most serious heavy metal pollutants in the coastal area and posed serious threats to marine organisms and human beings. Many studies have been conducted on its toxicological effects on living organisms from morphological
Wei Hseun Yeap et al.
Scientific reports, 6, 34310-34310 (2016-09-28)
Antibody-dependent cellular cytotoxicity (ADCC) is exerted by immune cells expressing surface Fcγ receptors (FcγRs) against cells coated with antibody, such as virus-infected or transformed cells. CD16, the FcγRIIIA, is essential for ADCC by NK cells, and is also expressed by
Richard O Francis et al.
The Journal of clinical investigation, 130(5), 2270-2285 (2020-01-22)
BACKGROUNDGlucose-6-phosphate dehydrogenase (G6PD) deficiency decreases the ability of red blood cells (RBCs) to withstand oxidative stress. Refrigerated storage of RBCs induces oxidative stress. We hypothesized that G6PD-deficient donor RBCs would have inferior storage quality for transfusion as compared with G6PD-normal
Judith M Myers et al.
Toxicology, 246(2-3), 222-233 (2008-03-11)
Hexavalent chromium [Cr(VI)] species such as chromates are cytotoxic. Inhalational exposure is a primary concern in many Cr-related industries and their immediate environments, and bronchial epithelial cells are directly exposed to inhaled Cr(VI). Chromates are readily taken up by cells
Laura Cristina Ramírez-Rodríguez et al.
Membranes, 10(12) (2020-12-04)
Water contamination by mercury and chromium has a direct effect in human health. A promising technology to remove heavy metals by membrane filtration is the use of hybrid membranes produced with whey protein fibrils (WPF) and activated carbon (AC). In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service