Skip to Content
Merck
All Photos(1)

Documents

287881

Sigma-Aldrich

Diphenylphosphine oxide

97%

Synonym(s):

HPOPh2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2P(O)H
CAS Number:
Molecular Weight:
202.19
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Hydrophosphonylations

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

mp

56-57 °C (lit.)

functional group

phosphine

SMILES string

O=[PH](c1ccccc1)c2ccccc2

InChI

1S/C12H11OP/c13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,14H

InChI key

ASUOLLHGALPRFK-UHFFFAOYSA-N

Application

Diphenylphosphine oxide can be used as a coupling partner to prepare various organophosphorus compounds via cross-coupling reaction with aryl halides in the presence of Ni/Zn catalyst.
It can also be used as a reactant to synthesize:
  • Diphenyl(2-phenyl-2H-indazol-3-yl)phosphine oxide derivatives by phosphonylation of 2H-indazoles using rose bengal as a catalyst.
  • Alkenyldiphenylphosphine oxides by hydrophosphinylation of terminal alkynes in the presence of transition metal catalysts.
  • Diphenylphosphino-containing chiral ligands for asymmetric catalytic reactions via Pd-catalyzed coupling reaction with aryl triflates.
  • Triarylphosphine oxides and Horner-Wittig reagents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile synthesis of chelating bisphosphine oxides and bisphosphines via palladium-catalyzed bishydrophosphinylation reactions
Allen Jr A, et al.
Tetrahedron Letters, 43(20), 3707-3710 (2002)
Visible-light-induced organophotoredox-catalyzed phosphonylation of 2 H-indazoles with diphenylphosphine oxide
Singsardar M, et al.
The Journal of Organic Chemistry, 83(20), 12694-12701 (2018)
Li-Biao Han et al.
Journal of the American Chemical Society, 126(16), 5080-5081 (2004-04-22)
The cheap nickel catalysts are more reactive than the corresponding noble metal catalysts in the catalytic additions of a variety of P(O)-H bonds and an S-H bond to alkynes, affording regio- and stereoselectively both the Markovnikov and the anti-Markovnikov adducts
Palladium-catalyzed air-based oxidative coupling of arylboronic acids with H-phosphine oxides leading to aryl phosphine oxides
Fu Tingting, et al.
Organic & Biomolecular Chemistry, 12(18), 2895-2902 (2014)
Alcock, N. W.; Brown, J. M.; Hulmes, D. I.
Tetrahedron Asymmetry, 4, 743-743 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service