Skip to Content
Merck
All Photos(2)

Key Documents

282278

Sigma-Aldrich

2,6-Difluorobenzamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3C(O)NH2
CAS Number:
Molecular Weight:
157.12
Beilstein:
2047480
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

145-148 °C (lit.)

solubility

ethanol: soluble 5%, clear to turbid, colorless to light yellow

SMILES string

NC(=O)c1c(F)cccc1F

InChI

1S/C7H5F2NO/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,(H2,10,11)

InChI key

AVRQBXVUUXHRMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Difluorobenzamide is a major metabolite of pesticide diflubenzuron and has been quantitated by HPLC/diode-array method.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Eriksson et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 30(10), 871-877 (1992-10-01)
The herbicides dichlobenil (2,6-dichlorobenzonitrile), chlorthiamid (2,6-dichlorothiobenzamide) and their environmental degradation product 2,6-dichlorobenzamide are irreversibly bound and toxic to the olfactory mucosa following single injections in mice (Brandt et al., Toxicology and Applied Pharmacology 1990, 103, 491-501; Brittebo et al., Fundamental
J Koerts et al.
Xenobiotica; the fate of foreign compounds in biological systems, 27(8), 801-817 (1997-08-01)
1. The metabolic fate of the insecticide teflubenzuron, orally dosed to the male Wistar rat, was investigated. Particular attention was paid to the metabolic fate of the benzoyl and aniline moiety after hydrolysis of the urea bridge. 2. The 0-48-h
HPLC/diode-array method for the determination of the pesticide diflubenzuron and its major metabolites 2,6-difluorobenzamide, 4-chlorophenylurea, and 4-chloroaniline in forestry matrixes.
Rodriguez E, et al.
Journal of Liquid Chromatography and Related Technologies, 21(12), 1857-1870 (1998)
Varahenage R Perera et al.
Biochimica et biophysica acta, 1840(9), 2851-2861 (2014-05-14)
Gram-positive bacteria in the phylum Firmicutes synthesize the low molecular weight thiol bacillithiol rather than glutathione or mycothiol. The bacillithiol transferase YfiT from Bacillus subtilis was identified as a new member of the recently discovered DinB/YfiT-like Superfamily. Based on structural

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service