Skip to Content
Merck
All Photos(3)

Key Documents

276375

Sigma-Aldrich

1,5-Naphthalenedisulfonic acid tetrahydrate

97%

Synonym(s):

Armstrong’s acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(SO3H)2·4H2O
CAS Number:
Molecular Weight:
360.36
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

SMILES string

O.O.O.O.OS(=O)(=O)c1cccc2c(cccc12)S(O)(=O)=O

InChI

1S/C10H8O6S2.4H2O/c11-17(12,13)9-5-1-3-7-8(9)4-2-6-10(7)18(14,15)16;;;;/h1-6H,(H,11,12,13)(H,14,15,16);4*1H2

InChI key

ZLBLYGIIADHDKG-UHFFFAOYSA-N

General description

1,5-Naphthalenedisulfonic acid tetrahydrate reacts with [n-Bu2SnO]n in a 1:1 stoichiometry followed by reaction with 2,2′-bipyridine-N,N′-dioxide (BPDO-I) to yield 1D-coordination polymer, [n-Bu2Sn(BPDO-I)(1,5-C10H6(SO3)2)]n.

Application



  • Assessment of verapamil in a myotonic dystrophy model: Research demonstrated verapamil′s efficacy in mitigating chloride and calcium channel dysfunctions in a myotonic dystrophy mouse model, suggesting its potential in treating related muscular dystrophies (Cisco et al., 2024).


  • Cardiac tissue modeling with verapamil: Verapamil hydrochloride was tested using human-induced pluripotent stem cell-derived cardiac tissues to evaluate drug interactions and cardiac responses, proving essential for advancing cardiac safety pharmacology (Miyagawa et al., 2024).




Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vadapalli Chandrasekhar et al.
Dalton transactions (Cambridge, England : 2003), 40(1), 114-123 (2010-11-19)
The reaction of [n-Bu(2)SnO](n) with 1,5-naphthalenedisulfonic acid tetrahydrate in a 1:1 stoichiometry followed by reaction with 2,2'-bipyridine-N,N'-dioxide (BPDO-I) afforded a 1D-coordination polymer [n-Bu(2)Sn(BPDO-I)(1,5-C(10)H(6)(SO(3))(2))](n) (1) where the disulfonate ligand acts as a bridging ligand between two tin centers. An analogous reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service