Skip to Content
Merck
All Photos(2)

Documents

270873

Sigma-Aldrich

2-Hydroxyquinoline

98%

Synonym(s):

2-Quinolinol, Carbostyril

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO
CAS Number:
Molecular Weight:
145.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

mp

198-199 °C (lit.)

solubility

alcohol: soluble(lit.)
diethyl ether: soluble(lit.)
dilute HCl: soluble(lit.)
water: slightly soluble (1 g/950mL)(lit.)

SMILES string

Oc1ccc2ccccc2n1

InChI

1S/C9H7NO/c11-9-6-5-7-3-1-2-4-8(7)10-9/h1-6H,(H,10,11)

InChI key

LISFMEBWQUVKPJ-UHFFFAOYSA-N

General description

2-Hydroxyquinoline is a specific inhibitor of plaque paraoxonase1 (PON1).

Application

2-Hydroxyquinoline was used to obtain the design of the platinum(IV) complexes with intense bands shifted towards longer wavelengths. The Pt(IV) complexes are inert stable prodrugs that were photoactivated to produce Pt(II) species with promising anticancer activity.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonios Keirouz et al.
Journal of nanobiotechnology, 18(1), 51-51 (2020-03-20)
The state-of-the-art hernia meshes, used in hospitals for hernia repair, are predominantly polymeric textile-based constructs that present high mechanical strength, but lack antimicrobial properties. Consequently, preventing bacterial colonization of implanted prosthetic meshes is of major clinical relevance for patients undergoing
Alexander J Bones et al.
Experimental parasitology, 196, 28-37 (2018-12-07)
Cryptosporidium is a genus of single celled parasites capable of infecting a wide range of animals including humans. Cryptosporidium species are members of the phylum apicomplexa, which includes well-known genera such as Plasmodium and Toxoplasma. Cryptosporidium parasites cause a severe
Shirin Biglari et al.
Microbial drug resistance (Larchmont, N.Y.), 23(5), 545-555 (2016-11-18)
Multidrug-resistant (MDR) Acinetobacter baumannii has increasingly emerged as an important nosocomial pathogen. The aim of this study was to determine the resistance profiles and genetic diversity in A. baumannii clinical isolates in a tertiary medical center in Malaysia. The minimum
Edyta Stec et al.
Chembiochem : a European journal of chemical biology, 12(11), 1724-1730 (2011-06-15)
Aurachins are quinoline alkaloids isolated from the myxobacterium Stigmatella aurantiaca. They are substituted with an isoprenoid side chain and act as potent inhibitors in the electron transport chain. A biosynthetic gene cluster that contains at least five genes (auaA-auaE) has
Arianna Friggeri et al.
Journal of controlled release : official journal of the Controlled Release Society, 97(2), 241-248 (2004-06-16)
Gels of low molecular weight gelators (LMWGs) are self-assembled, thermoreversible, viscoelastic materials which can also be rendered sensitive to light, pH or chemical substances by simple chemical modifications. In addition, the ability of some of these LMWGs to gelate water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service