Skip to Content
Merck
All Photos(2)

Documents

245771

Sigma-Aldrich

Isobutyric anhydride

97%

Synonym(s):

2-Methylpropionic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2COCH(CH3)2
CAS Number:
Molecular Weight:
158.19
Beilstein:
386267
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.45 (vs air)

vapor pressure

10 mmHg ( 67 °C)

Assay

97%

form

liquid

autoignition temp.

624 °F

expl. lim.

1.09 %, 87 °F
7.7 %, 127 °F

refractive index

n20/D 1.406 (lit.)

bp

182 °C (lit.)

mp

−56 °C (lit.)

density

0.954 g/mL at 25 °C (lit.)

SMILES string

CC(C)C(=O)OC(=O)C(C)C

InChI

1S/C8H14O3/c1-5(2)7(9)11-8(10)6(3)4/h5-6H,1-4H3

InChI key

LSACYLWPPQLVSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The mechanism of esterification of 1-(1-naphthyl)ethanol by isobutyric anhydride with 4-pyrrolidinopyridine as catalyst was studied.

Application

Isobutyric anhydride was used in the synthesis of 4-O-isobutyryl derivative via reaction with octyl β-D-glucopyranoside in the presence of C2-symmetric chiral 4-pyrrolidinopyridine as a catalyst.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin Wang et al.
Chemistry, an Asian journal, 10(8), 1690-1697 (2015-06-03)
Hyperbranched polyethylenimine terminated with isobutyramide groups (HPEI-IBAm), 4-(phenylazo)benzoic acid (PABA), and α-cyclodextrin (α-CD) were assembled together at pH≈7 to form the three-component supramolecular complexes that were verified by (1) H and 2D ROESY (1) H NMR spectroscopy. UV/Vis spectrometric titration experiments
Takeo Kawabata et al.
Journal of the American Chemical Society, 129(42), 12890-12895 (2007-10-02)
An organocatalytic method for the chemo- and regioselective acylation of monosaccharides has been developed. Treatment of octyl beta-D-glucopyranoside with isobutyric anhydride in the presence of 10 mol % of a C2-symmetric chiral 4-pyrrolidinopyridine catalyst (1) at -50 degrees C gave
Evgeny Larionov et al.
Journal of the American Chemical Society, 134(22), 9390-9399 (2012-05-10)
The mechanism of esterification of the secondary alcohol 1-(1-naphthyl)ethanol 9 by isobutyric anhydride catalyzed by 4-pyrrolidinopyridine (PPY, 11) and a series of single enantiomer atropisomeric 4-dialkylaminopyridines 8a-g has been studied computationally at the B3LYP/6-311+G(d,p)//B3LYP/6-31G(d) level. Comparison of the levels of
Xiaoning Shan et al.
International journal of pharmaceutics, 590, 119884-119884 (2020-09-21)
Poly(2-methyl-2-oxazoline) (PMOZ), poly(2-propyl-2-oxazoline) (PnPOZ) and poly(2-isopropyl-2-oxazoline) (PiPOZ) were synthesized by hydrolysis of 50 kDa poly(2-ethyl-2-oxazoline) (PEOZ) and subsequent reaction of the resulting poly(ethylene imine) with acetic, butyric and isobutyric anhydrides, respectively. These polymers were characterized by proton nuclear magnetic resonance, FTIR
Rosamaria Capuano et al.
Bio-protocol, 7(23), e2642-e2642 (2017-12-05)
Human induced pluripotent stem cells (hiPSCs) are a promising tool in cell-based therapies for degenerative diseases. A safe application of hiPSCs in vivo, requires the detection of the presence of residual undifferentiated pluripotent cells that can potentially cause the insurgence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service