Skip to Content
Merck
All Photos(4)

Key Documents

241911

Sigma-Aldrich

Tungsten(VI) chloride

≥99.9% trace metals basis

Synonym(s):

Tungsten hexachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
WCl6
CAS Number:
Molecular Weight:
396.56
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.9% trace metals basis
form:
powder

vapor pressure

43 mmHg ( 215 °C)

Quality Level

Assay

≥99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: tungsten

impurities

≤1000.0 ppm Trace Metal Analysis

bp

347 °C (lit.)

mp

275 °C (lit.)

density

3.52 g/mL at 25 °C (lit.)

SMILES string

Cl[W](Cl)(Cl)(Cl)(Cl)Cl

InChI

1S/6ClH.W/h6*1H;/q;;;;;;+6/p-6

InChI key

KPGXUAIFQMJJFB-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

General description

Tungsten(VI) chloride is a highly reactive crystalline solid commonly used in the field of catalysis, perovskite solar cells, and light-emitting devices. It is a diamagnetic solid. However, it can be highly corrosive and have strong oxidizing effects.

Application

Tungsten(VI) chloride can be used:
  • As a starting material to synthesize tungsten nanoparticles and Mo-doped urchin-like W18O49 Nanostructure using the hydrothermal method. The Mo-W18O49 electrocatalyst exhibited excellent electrocatalytic activity toward Hydrogen Evolution Reaction (HER). By doping Mo species into defect-rich W18O49 ultrathin nanowires, it has also been demonstrated to be an excellent candidate for photocatalytic N2 fixation to ammonia.
  • To synthesize crystalline mesoporous WO3 with 11 nm pore size utilizing a high-molecular-weight amphiphilic block copolymer as the structure-directing agent. The materials performed admirably in terms of H2S gas sensing.
  • To fabricate Tungsten disulfide and WS2/reduced graphene oxide (WS2/rGO) nanosheets by hydrothermal synthesis. The WS2/rGO nanosheets showed exceptional electrocatalytic activity for the hydrogen evolution reaction.
  • To produce the WS2-nanoflowers@rGO and nitrogen-doped carbon spheres@WS2 composite as an anode material for enhanced electrode performance in lithium-ion batteries.
  • As a dopant to fabricate TiO2 compact layers for perovskite solar cells with enhanced performance.
  • As a catalyst to prepare self-healing epoxy composites with microcapsules.
  • As a catalyst for transamidation of tertiary alkyl amides.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hierarchical Nanocomposite of Hollow N-Doped Carbon Spheres Decorated with Ultrathin WS2 Nanosheets for High-Performance Lithium-Ion Battery Anode
Xiaohui Zeng, et al.
ACS Applied Materials & Interfaces, 8, 18841-18848 (2016)
Giuseppe Bengasi et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(35), 8313-8320 (2019-04-03)
Oxidative chemical vapour deposition of (5,15-diphenylporphyrinato)nickel(II) (NiDPP) with iron(III) chloride as oxidant yielded a conjugated poly(metalloporphyrin) as a highly coloured thin film, which is potentially useful for optoelectronic applications. This study clarified the reactive sites of the porphyrin monomer NiDPP
Liyan Zhou et al.
Scientific reports, 9(1), 1357-1357 (2019-02-06)
Tungsten Disulfide (WS2) is considered to be a promising Hydrogen Evolution Reaction (HER) catalyst to replace noble metals (such as Pt and Pd). However, progress in WS2 research has been impeded by the inertness of the in-plane atoms during HER.
Refining Defect States in W18O49 by Mo Doping: A Strategy for Tuning N2 Activation towards Solar-Driven Nitrogen Fixation
Ning Zhang, et al
Journal of the American Chemical Society, 140, 9434-9443 (2018)
Preparation and self-healing performance of epoxy composites with microcapsules and tungsten (VI) chloride catalyst
Li Haiyan, et al.
Journal of Reinforced Plastics and Composites, 31, 924-932 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service