Skip to Content
Merck
All Photos(1)

Documents

236349

Sigma-Aldrich

Butyryl chloride

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COCl
CAS Number:
Molecular Weight:
106.55
Beilstein:
605395
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.7 (vs air)

Assay

≥99%

form

liquid

refractive index

n20/D 1.412 (lit.)

bp

102 °C (lit.)

mp

−89 °C (lit.)

solubility

diethyl ether: miscible(lit.)

density

1.026 g/mL at 25 °C (lit.)

SMILES string

CCCC(Cl)=O

InChI

1S/C4H7ClO/c1-2-3-4(5)6/h2-3H2,1H3

InChI key

DVECBJCOGJRVPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Photchlorination of butyryl chloride in liquid phase has been investigated.

Application

Butyryl chloride can be used as:
  • A reagent in the acylation of thiophene in the liquid phase catalyzed by zeolites.
  • A source of CO and -Cl in the chlorocarbonylation of aryl bromides to yield acid chlorides.
  • An intermediate in the synthesis of aryl [11C]methylsulfones for PET applications.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

64.4 °F - DIN 51755 Part 1

Flash Point(C)

18 °C - DIN 51755 Part 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CHLORINATION OF BUTYRYL CHLORIDE IN THE LIQUID-PHASE.
Korhonen IO.
Acta Chemica Scandinavica. Series B, 35(3), 175-178 (1981)
A lewis acid site-activated reaction in zeolites: thiophene acylation by butyryl chloride.
Isaev Y and Fripiat JJ.
J. Catal., 182(1), 257-263 (1999)
A general method for the synthesis of aryl [11C] methylsulfones: potential PET probes for imaging cyclooxygenase-2 expression
Majo VJ, et al.
Bioorganic & Medicinal Chemistry Letters, 15(19), 4268-4271 (2005)
A Lewis acid site-activated reaction in zeolites: Thiophene acylation by butyryl chloride
Isaev Y and Fripiat JJ
J. Catal., 182(1), 257-263 (1999)
Palladium-catalyzed Chlorocarbonylation of Aryl (pseudo) Halides through in situ Generation of Carbon Monoxide
Boehm P, et al.
Angewandte Chemie (International Edition in English), 132(41), 18043-18052 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service