Skip to Content
Merck
All Photos(2)

Key Documents

236152

Sigma-Aldrich

Lead(II) fluoride

powder, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbF2
CAS Number:
Molecular Weight:
245.20
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99%
form:
powder

Quality Level

Assay

≥99%

form

powder

reaction suitability

reagent type: catalyst
core: lead

mp

824 °C (lit.)

density

8.445 g/mL at 25 °C (lit.)

SMILES string

F[PbH2]F

InChI

1S/2FH.Pb/h2*1H;/q;;+2/p-2

InChI key

FPHIOHCCQGUGKU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PbF2 compared to Al2O3 and AlF3 to produce an epithermal neutron beam for radiotherapy.
H B Liu
Medical physics, 23(2), 279-280 (1996-02-01)
C M Tabchoury et al.
Oral diseases, 5(2), 100-103 (1999-10-16)
Results from previous studies have shown that pre- and perinatal exposure to lead enhances susceptibility of rats to development of dental caries. A possible explanation for this phenomenon may be that lead complexes with fluoride and renders F insoluble and
Alicia Glatfelter et al.
The Analyst, 131(11), 1207-1209 (2006-10-27)
We demonstrate a multi-spectrum technique for facile, quantitative determination of lead in solid materials using solid-state (207)Pb NMR that avoids the major problem of uniform excitation across a wide spectral range; the method can be employed without chemical separation or
A S Filin et al.
Gigiena i sanitariia, (3)(3), 28-29 (1992-03-01)
Effect of lead fluoride dust on the micronuclear reticulocyte content in the bone marrow was shown. Some functional changes in the testicles and increase in the fetal death rate in exposure to 50 mg/m3 of lead fluoride dust were noted.
A I Fadeev
Meditsina truda i promyshlennaia ekologiia, (10)(10), 4-6 (1996-01-01)
The article deals with results obtained in hygienic evaluation of production and application of new monocrystalline compounds based on cesium, sodium, iodine, lead, scandium, fluorine. The evaluation covered work conditions and health state of those engaged into production of monocrystalline

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service