Skip to Content
Merck
All Photos(3)

Key Documents

219096

Sigma-Aldrich

D-Aspartic acid

99%, for peptide synthesis, ReagentPlus®

Synonym(s):

(R)-(−)-Aminosuccinic acid, (R)-2-Aminosuccinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
133.10
Beilstein:
1723529
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Product Name

D-Aspartic acid, ReagentPlus®, 99%

Quality Level

product line

ReagentPlus®

Assay

99%

form

powder

optical activity

[α]20/D −24°, c = 2.3 in 6 M HCl

optical purity

ee: 98% (GLC)

reaction suitability

reaction type: solution phase peptide synthesis

mp

>300 °C (lit.)

application(s)

peptide synthesis

SMILES string

N[C@H](CC(O)=O)C(O)=O

InChI

1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/t2-/m1/s1

InChI key

CKLJMWTZIZZHCS-UWTATZPHSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application


  • Metabolomics Analysis Identifies Differential Metabolites as Biomarkers for Acute Myocardial Infarction.: This study utilizes metabolomics to identify biomarkers for acute myocardial infarction, showcasing D-Aspartic acid′s role in amino acid metabolism and potential diagnostic applications. (Zhou et al., 2024).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco Errico et al.
Amino acids, 43(5), 1861-1871 (2012-08-02)
Free D-aspartate (D-Asp) occurs in substantial amounts in the brain at the embryonic phase and in the first few postnatal days, and strongly decreases in adulthood. Temporal reduction of D-Asp levels depends on the postnatal onset of D-aspartate oxidase (DDO)
Akifumi Oda et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(29), 3337-3343 (2011-08-30)
Molecular dynamics simulations of amyloid β(1-42) containing D-aspartic acid residues were performed using several continuous solvent models to investigate the usefulness of simulation methods for D-amino acid-containing proteins and peptides. Normal molecular dynamics simulations and replica exchange molecular dynamics simulations
Antimo D'Aniello
Brain research reviews, 53(2), 215-234 (2006-11-23)
D-Aspartic acid (d-Asp), an endogenous amino acid present in vertebrates and invertebrates, plays an important role in the neuroendocrine system, as well as in the development of the nervous system. During the embryonic stage of birds and the early postnatal
Ikuhiko Noji et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(29), 3310-3316 (2011-07-12)
Unusual amino acid residues such as L-β-aspartyl (Asp), D-α-Asp, and D-β-Asp have been detected in proteins and peptides such as α-crystallin in the lens and β-amyloid in the brain. These residues increase with age, and hence they are associated with
H Homma
Amino acids, 32(1), 3-11 (2006-06-07)
Recent investigations have shown that D-aspartate (D-Asp) plays an important physiological role(s) in the mammalian body. Here, several recent studies of free D-Asp metabolism in mammals, focusing on cellular localization in tissues, intracellular localization, biosynthesis, efflux, uptake and degradation are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service