Skip to Content
Merck
All Photos(2)

Documents

196657

Sigma-Aldrich

1-Fluoronaphthalene

99%

Synonym(s):

α-Fluoronaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7F
CAS Number:
Molecular Weight:
146.16
Beilstein:
1906413
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.593 (lit.)

bp

215 °C (lit.)

mp

−13 °C (lit.)

density

1.1322 g/mL at 20 °C (lit.)

SMILES string

Fc1cccc2ccccc12

InChI

1S/C10H7F/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H

InChI key

CWLKTJOTWITYSI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flash photolysis of O2 and 1-fluoronaphthalene mixtures in the gas phase have been investigated.

Application

1-Fluoronaphthalene was used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It was also used in the synthesis of LY248686, a potent inhibitor of serotonin and norepinephrine uptake.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Pawlas et al.
Organic letters, 4(16), 2687-2690 (2002-08-03)
[reaction: see text] A one-pot, t-BuLi-induced synthesis of 6-substituted phenanthridines from fluoroarenes and nitriles via 1,2-arynes is reported. Aryl- and hetaryl nitriles, cyanamides, and trimethylacetonitrile gave phenanthridine products. The method was extended to provide bisphenanthridine 10 by a one-pot bis-cyclization
Formation of O 2 (1Sigma g+) by 1-fluoronaphthalene sensitization.
Andrews LJ and Abrahamson EW.
Chemical Physics Letters, 10, 113-116 (1971)
Asymmetric synthesis and absolute stereochemistry of LY248686.
Deeter J, et al.
Tetrahedron Letters, 31(49), 7101-7104 (1990)
Peter Wipf et al.
Organic letters, 5(7), 1155-1158 (2003-03-28)
[reaction: see text] Electron-rich dinaphthyl ethers were synthesized by S(N)Ar reactions between naphthols and activated fluoronaphthalenes. 2-tert-Butyl-1,1,3,3-tetramethylguanidine (Barton's base) was found to be an excellent, mild alternative to traditional inorganic bases for promoting the coupling reaction.
C E Cerniglia et al.
Applied and environmental microbiology, 48(2), 294-300 (1984-08-01)
The metabolism of 1-fluoronaphthalene by Cunninghamella elegans ATCC 36112 was studied. The metabolites were isolated by reverse-phase high-pressure liquid chromatography and characterized by the application of UV absorption, 1H nuclear magnetic resonance, and mass spectral techniques. C. elegans oxidized 1-fluoronaphthalene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service