Skip to Content
Merck
All Photos(2)

Key Documents

182249

Sigma-Aldrich

Poly(methyl methacrylate-co-ethyl acrylate)

ethyl acrylate <5 wt. %, average Mn ~39,500 by GPC, average Mw ~101,000 by GPC, powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)(CO2CH3)]X[CH2CH(CO2C2H5]Y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

autoignition temp.

580 °F

mol wt

average Mn ~39,500 by GPC
average Mw ~101,000 by GPC

composition

ethyl acrylate, <5 wt. %

transition temp

Tg (DSC) 104 °C (onset)

SMILES string

O(CC)C(=O)C=C.O(C)C(=O)C(=C)C

InChI

1S/2C5H8O2/c1-4(2)5(6)7-3;1-3-5(6)7-4-2/h1H2,2-3H3;3H,1,4H2,2H3

InChI key

XPNLOZNCOBKRNJ-UHFFFAOYSA-N

Application

Poly(methyl methacrylate-co-ethyl acrylate) (PMMAEA) copolymers may be used to prepare single phase mixture with phenolic derivative. PMMAEA may be used to form latex interpenetrating polymer networks (LIPNs). Membranes of PMMAEA were reportedly used for separation of heavy metal ions from aqueous solutions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Horisawa et al.
Drug development and industrial pharmacy, 26(12), 1271-1278 (2001-01-09)
Some acidic nonsteroidal anti-inflammatory drugs (NSAIDs) are poorly soluble in the stomach. In this study, M-5011, d-2-[4-(3-methyl-2-thienyl) phenyl] propionic acid, was used as a model substance. To increase the dissolution rate of M-5011, a solid dispersion of M-5011 was prepared
A Y Lin et al.
AAPS pharmSci, 3(2), E14-E14 (2001-12-14)
This study investigates the crystallization of the endogenous surfactant nonoxynol 100 in Eudragit NE30D-free films during storage and the influences of nonoxynol 100 on the dissolution of diphenhydramine hydrochloric acid (HCl) pellets coated with Eudragit NE30D before and after aging
Structure development in the phenolic resin/poly (methyl methacrylate?co?ethyl acrylate) copolymer blends.
Kim B S, et al.
Journal of Applied Polymer Science, 68(11), 1829-1837 (1998)
S V Lafferty et al.
International journal of pharmaceutics, 235(1-2), 107-111 (2002-03-07)
Dynamic Mechanical Thermal Analysis of cast and sprayed films of an aqueous dispersion of polymethyl methacrylate (Eudragit NE30D) and mixtures with an aqueous dispersion of ethylcellulose (Aquacoat ECD-30) has been undertaken. Such analysis allows the identification of glass transition temperatures
n?Butyl acrylate based latex interpenetrating polymer networks used as processing modifiers and impact modifiers of poly (vinyl chloride).
Zhang L, et al.
Journal of Applied Polymer Science, 91(2), 1168-1173 (2004)

Articles

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service