177466
4-Methylcyclohexylamine, mixture of cis and trans
97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3C6H10NH2
CAS Number:
Molecular Weight:
113.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
liquid
refractive index
n20/D 1.4531 (lit.)
bp
151-154 °C (lit.)
density
0.855 g/mL at 25 °C (lit.)
SMILES string
CC1CCC(N)CC1
InChI
1S/C7H15N/c1-6-2-4-7(8)5-3-6/h6-7H,2-5,8H2,1H3
InChI key
KSMVBYPXNKCPAJ-UHFFFAOYSA-N
General description
Trans-4-methylcyclohexylamine is a spermidine synthase inhibitor.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
80.6 °F - closed cup
Flash Point(C)
27 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Masaki Kobayashi et al.
Biological & pharmaceutical bulletin, 29(5), 863-867 (2006-05-03)
A close relationship between rat liver regeneration and the concentration ratio of spermidine to spermine (spd:spm) was demonstrated by the oral administration of trans-4-methylcyclohexylamine (MCHA), a specific inhibitor of putrescine aminopropyltransferase. A decrease in recovery rate of remnant liver with
Keijiro Samejima et al.
Biological & pharmaceutical bulletin, 30(10), 1943-1946 (2007-10-06)
A sensitive method for the determination of polyamines in mammalian cells was described using electrospray ionization and time-of-flight mass spectrometer. This method was 50-fold more sensitive than the previous method using ionspray ionization and quadrupole mass spectrometer. The method employed
Trans-4-methylcyclohexylamine, a potent new inhibitor of spermidine synthase.
A Shirahata et al.
Chemical & pharmaceutical bulletin, 36(8), 3220-3222 (1988-08-01)
Keijiro Samejima
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 126(8), 529-542 (2006-08-02)
This review describes my work in the field of polyamine research for the last 35 years. My research started with developing the improved synthesis of decarboxylated S-adenosylmethionine and then moved to the purification of spermidine synthase from rat prostate. I
TachaZi Plym Forshell et al.
Cancer prevention research (Philadelphia, Pa.), 3(2), 140-147 (2010-01-28)
The oncogenic transcription factor c-Myc (Myc) is frequently overexpressed in human cancers. Myc is known to induce or repress a large set of genes involved in cell growth and proliferation, explaining the selection for mutations in cancer that deregulate Myc
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service