Skip to Content
Merck
All Photos(3)

Documents

135232

Sigma-Aldrich

Hydrocinnamic acid

99%

Synonym(s):

3-Phenylpropionic acid, Benzylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2COOH
CAS Number:
Molecular Weight:
150.17
Beilstein:
907515
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

280 °C (lit.)

mp

45-48 °C (lit.)

solubility

ethanol: soluble 50 mg/mL, clear, colorless to faintly yellow

density

1.071 g/mL at 25 °C (lit.)

SMILES string

OC(=O)CCc1ccccc1

InChI

1S/C9H10O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,10,11)

InChI key

XMIIGOLPHOKFCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrocinnamic acid forms complexes with uranium(VI) and has been investigated in aqueous solution by attenuated total reflection fourier transform infrared spectroscopy. Hydrocinnamic acids are the major rhizospheric compounds with known growth regulatory activity.

Application

Hydrocinnamic acid was used in the synthesis of three medium-chain acyl-Coenzyme A′s i.e. 3-phenylpropionyl-CoA from mixed anhydrides of fatty acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hany F Sobhi et al.
Analytical biochemistry, 401(1), 114-124 (2010-02-27)
The measurement of acyl-CoA dehydrogenase activities is an essential part of the investigation of patients with suspected defects in fatty acid oxidation. Multiple methods are available for the synthesis of the substrates used for measuring acyl-CoA dehydrogenase activities; however, the
C S Tang et al.
Plant physiology, 69(1), 155-160 (1982-01-01)
Collection of allelopathic chemicals from the undisturbed plant root system is difficult because of their low concentrations and the high level of contaminants in growth media such as soil. A new approach for the continuous trapping of quantities of extracellular
Astrid Barkleit et al.
Applied spectroscopy, 62(7), 798-802 (2008-10-22)
Uranyl complexes with phenylalanine and the analogous ligand phenylpropionate were investigated in aqueous solution by attenuated total reflection (ATR) Fourier transform infrared (FT-IR) spectroscopy. The assignment of the observed bands to vibrational modes was accomplished using spectra of the pure
Nuria Mateo Anson et al.
Journal of agricultural and food chemistry, 57(14), 6148-6155 (2009-06-23)
Ferulic acid (FA) is the most abundant phenolic compound in wheat grain, mainly located in the bran. However, its bioaccessibility from the bran matrix is extremely low. Different bioprocessing techniques involving fermentation or enzymatic and fermentation treatments of wheat bran
Ling Cheng et al.
American journal of physiology. Gastrointestinal and liver physiology, 290(6), G1307-G1317 (2006-01-28)
Platelet-activating factor (PAF) and interleukin-6 (IL-6) are produced in the esophagus in response to HCl and affect ACh release, causing changes in esophageal motor function similar to esophagitis (Cheng L, Cao W, Fiocchi C, Behar J, Biancani P, and Harnett

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service