Skip to Content
Merck
All Photos(1)

Key Documents

134716

Sigma-Aldrich

3-Butenoic acid

97%

Synonym(s):

Vinylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2COOH
CAS Number:
Molecular Weight:
86.09
Beilstein:
1699159
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

contains

hydroquinone as inhibitor

refractive index

n20/D 1.423 (lit.)

bp

163 °C (lit.)

mp

−39 °C (lit.)

density

1.013 g/mL at 25 °C (lit.)

functional group

allyl
carboxylic acid

storage temp.

2-8°C

SMILES string

OC(=O)CC=C

InChI

1S/C4H6O2/c1-2-3-4(5)6/h2H,1,3H2,(H,5,6)

InChI key

PVEOYINWKBTPIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Butenoic acid was used in the synthesis of polyethylene glycolated boltorn coatings[1]. It was used to study the inhibition of the enzyme-catalysed conversion of D-tyrosyl- phenylalanyl-glycine to D-tyrosyl-phenylaline amide by non-peptide carboxylic acids[2]. It was used in preparation of strained nitroso Diels-Aldrer adducts which undergo a domino ROC metathesis in the presence of Ru-carbene catalysts and olefins[3].

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos Adelantado et al.
Electrophoresis, 39(12), 1437-1442 (2018-03-27)
It is described a simple, rapid and efficient methodology to characterise and separate gold nanoprisms and nanooctahedra by capillary electrophoresis. This technique is suitable to distinguish between morphologies and it can be used as a powerful separation tool after a
Géraldine Calvet et al.
Organic letters, 9(8), 1485-1488 (2007-03-14)
[reaction: see text] Strained bicyclic 3,6-dihydro-1,2-oxazine is a reactive substrate in domino metathesis with an external alkene. This general transformation leads to isoxazolo[2,3-a]pyridin-7-one, a versatile scaffold combining chemical functions that could be selectively oxidized or reduced. The synthetic relevance of
Xiao-Ding Xu et al.
Colloids and surfaces. B, Biointerfaces, 59(2), 158-163 (2007-06-20)
To investigate the temperature sensitive mechanism and develop temperature sensitive materials, a novel poly(N-isopropyl-3-butenamide) (PNIPBAm) hydrogel was designed and synthesized. The maximum swelling ratio, temperature dependence of swelling ratio and deswelling/reswelling kinetics of the resulted PNIPBAm hydrogel were characterized. The
Meghan E Dunn et al.
The journal of physical chemistry. A, 112(41), 10226-10235 (2008-09-19)
In this study we present the gas-phase vibrational spectrum of vinylacetic acid with a focus on the nu = 1-5 vibrational states of the OH stretching transitions. Cross sections for nu = 1, 2, 4 and 5 of the OH
Lvfeng Zhu et al.
The Journal of organic chemistry, 75(17), 6027-6030 (2010-08-13)
A challenging direct asymmetric hydrogenation of (E)-2-oxo-4-arylbut-3-enoic acids to give 2-hydroxy-4-arylbutanoic acids (85.4-91.8% ee) was achieved with a Ru catalyst based on SunPhos as the chiral ligand. Further investigation of the reaction revealed that partial isomerization of 2-hydroxy-4-arylbutenoic acids was

Questions

  1. Is this 3-Butenoic acid a solution or a solid?

    1 answer
    1. This product is a neat liquid with a density of 1.013 g/mL at 25 °C.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service