Skip to Content
Merck
All Photos(1)

Documents

130532

Sigma-Aldrich

5,7-Dichloro-8-hydroxy-2-methylquinoline

98%

Synonym(s):

5,7-Dichloro-2-methyl-8-quinolinol, 5,7-Dichloro-8-hydroxyquinaldine, 5,7-Dichloro-8-quinaldinol, BCM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7Cl2NO
CAS Number:
Molecular Weight:
228.07
Beilstein:
156683
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

108-112 °C (dec.) (lit.)

SMILES string

Cc1ccc2c(Cl)cc(Cl)c(O)c2n1

InChI

1S/C10H7Cl2NO/c1-5-2-3-6-7(11)4-8(12)10(14)9(6)13-5/h2-4,14H,1H3

InChI key

GPTXWRGISTZRIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Therapeutic efficacy of the combination of chlorquinaldol plus promestriene in infections of the vagina].
S Palacios Gil-Antuñano
Revista clinica espanola, 173(5-6), 297-300 (1984-06-15)
Cutaneous drug reaction case reports: from the world literature.
American journal of clinical dermatology, 3(1), 63-69 (2002-01-31)
Contact cross-sensitization among quinolines.
A Rodríguez et al.
Allergy, 56(8), 795-795 (2001-08-08)
A Dimitrova et al.
Akusherstvo i ginekologiia, 46(1), 46-47 (2007-05-02)
The data of 30 nonpregnant women in reproductive age BV treated with Gynalgin were presented. On clinical and microbiological indicator Gynalgin showed a good effect on BV-associated microorganisms (80%). Only one of the cases of BV was not affected (4%).
Allergic contact dermatitis due to benzoyl peroxide in an arm prosthesis.
C Vincenzi et al.
Contact dermatitis, 24(1), 66-67 (1991-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service