Skip to Content
Merck
All Photos(2)

Key Documents

124400

Sigma-Aldrich

Vinyl pivalate

contains hydroquinone as inhibitor, 99%

Synonym(s):

Vinyl trimethylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCO2CH=CH2
CAS Number:
Molecular Weight:
128.17
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

contains

hydroquinone as inhibitor
5-30 ppm Hydroquinone (HQ) as inhibitor
~10 ppm Hydroquinone as inhibitor (CAS 123-31-9)

refractive index

n20/D 1.405 (lit.)

bp

110 °C (lit.)

density

0.866 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)C(=O)OC=C

InChI

1S/C7H12O2/c1-5-9-6(8)7(2,3)4/h5H,1H2,2-4H3

InChI key

YCUBDDIKWLELPD-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Carc. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eunae Cho et al.
Scientific reports, 6, 23740-23740 (2016-03-30)
Amphiphilic cyclodextrins have been synthesized with self-acylating reaction using vinyl esters in dimethylformamide. In the present study no base, catalyst, or enzyme was used, and the structural analyses using thin layer chromatography, nuclear magnetic resonance spectroscopy and mass spectrometry show
Mengmeng Peng et al.
Environmental pollution (Barking, Essex : 1987), 259, 113854-113854 (2020-01-10)
The main objectives of the present study were to investigate urinary monohydroxylated polycyclic aromatic hydrocarbons (OH-PAHs) in 77 primiparas who live in Shenzhen, Guangdong Province, China, and their association with 8-hydroxy-2'-deoxyguanosine (8-OHdG) and human health risks. High detection frequencies of
Yingxin Yu et al.
Journal of hazardous materials, 401, 123390-123390 (2020-07-14)
Monohydroxylated polycyclic aromatic hydrocarbons (OH-PAHs), phthalate metabolites (mPAEs), and 8-hydroxy-2'-deoxyguanosine (8-OHdG) in the urine of school children aged 8-11 years from Shenzhen, China were measured in order to investigate oxidative stress damage from co-exposure to PAHs and PAEs. The concentrations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service