Skip to Content
Merck
All Photos(1)

Key Documents

121967

Sigma-Aldrich

Aminoacetaldehyde dimethyl acetal

99%

Synonym(s):

2,2-Dimethoxyethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH(OCH3)2
CAS Number:
Molecular Weight:
105.14
Beilstein:
741868
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.417 (lit.)

bp

135-139 °C/95 mmHg (lit.)

density

0.965 g/mL at 25 °C (lit.)

functional group

acetal
amine
ether

SMILES string

COC(CN)OC

InChI

1S/C4H11NO2/c1-6-4(3-5)7-2/h4H,3,5H2,1-2H3

InChI key

QKWWDTYDYOFRJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aminoacetaldehyde dimethyl acetal reacts with sulfone, followed by hydrolysis and reductive amination by adding desired piperazine derivative to yield piperazine derivatives of 2-furanyl[1,2,4]triazolo[1,5-a][1,3,5]triazine.

Aminoacetaldehyde dimethyl acetal is used as a building block for the synthesis of various acylated and sufonylated oxyenamides.

Application

Aminoacetaldehyde dimethyl acetal was used in a study to develop a fluorescent substrate for aldehyde dehydrogenase. It was used in preparation of chitosan-dendrimer hybrids having various functional groups such as carboxyl, ester and poly(ethylene glycol). It was used in an efficient 3-step synthesis of a bicyclic proline analog from L-ascorbic acid and in 3-component reaction catalyzed by MgClO4 leading to α-aminophosphonates.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved synthesis of imidazole-2-carboxaldehyde, imidazole-2-carboxylic acid, and ethyl imidazole-2-carboxylate
Galeazzi, Eduviges and Guzman
The Journal of Organic Chemistry, 60, 1090-1092 (1995)
Stereoselective Synthesis of 2-Oxyenamides
Krieg, Sara-Cathrin and Grimmer
European Journal of Organic Chemistry (2022)
R W Storms et al.
Proceedings of the National Academy of Sciences of the United States of America, 96(16), 9118-9123 (1999-08-04)
Because hematopoietic stem cells are rich in aldehyde dehydrogenase (ALDH) activity, we developed a fluorescent substrate for ALDH, termed BODIPY aminoacetaldehyde (BAAA), and tested its potential for isolating primitive human hematopoietic cells. A population of cells with low orthogonal light
Hitoshi Sashiwa et al.
Biomacromolecules, 4(5), 1244-1249 (2003-09-10)
Chitosan-dendrimer hybrids having various functional groups such as carboxyl, ester, and poly(ethylene glycol) groups were prepared successfully using dendrimer acetal by reductive N-alkylation. The synthetic procedure could be accomplished by one-step reaction without organic solvent. The degree of substitution of
Chi B Vu et al.
Bioorganic & medicinal chemistry letters, 14(19), 4835-4838 (2004-09-03)
Piperazine derivatives of 2-furanyl[1,2,4]triazolo[1,5-a][1,3,5]triazine have recently been shown to be potent and selective adenosine A(2a) receptor antagonists. We now demonstrate that potent and selective A(2a) receptor antagonists could still be obtained when the arylpiperazines are separated from the triazolotriazine core

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service