Skip to Content
Merck
All Photos(2)

Key Documents

113018

Sigma-Aldrich

Crotonic acid

98%

Synonym(s):

trans-2-Butenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCOOH
CAS Number:
Molecular Weight:
86.09
Beilstein:
1719943
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.97 (vs air)

Quality Level

vapor pressure

0.19 mmHg ( 20 °C)

Assay

98%

autoignition temp.

744 °F

expl. lim.

15.1 %, 171 °F
2.2 %, 135 °F

bp

180-181 °C (lit.)

mp

70-72 °C (lit.)

solubility

H2O: soluble

density

1.027 g/mL at 25 °C (lit.)

SMILES string

[H]\C(C)=C(\[H])C(O)=O

InChI

1S/C4H6O2/c1-2-3-4(5)6/h2-3H,1H3,(H,5,6)/b3-2+

InChI key

LDHQCZJRKDOVOX-NSCUHMNNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Crotonic acid undergoes esterification with butan-2-ol by following second order reaction kinetics.

Application

Crotonic acid is used in the copolymerization of crotonic acid hydrogel systems by using gamma-rays. The crosslinked copolymeric crotonic acid hydrogels release fertilizers and drugs that help to prevent environmental pollution.

Preparation Note

Crotonic acid dissolves in water at a concentartion of 100 mg/ml to yield a clear, colorless solution.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Donya Kamravamanesh et al.
AMB Express, 7(1), 143-143 (2017-07-09)
Poly-β-hydroxybutyrate (PHB) production from CO
R F Bing et al.
Hypertension (Dallas, Tex. : 1979), 5(6), 951-957 (1983-11-01)
We have studied the possible vasodepressor role of the renal medulla by chemical medullectomy. Bromoethylamine hydrobromide (200 mg/kg) was injected to induce selective renal medullary necrosis in rats. The acute effects on sodium balance and long-term effects on blood pressure
Rita Mozes-Koch et al.
Journal of biological engineering, 11, 44-44 (2017-12-05)
Previously we demonstrated that an entire bacterial operon (the PRN operon) is expressible in plants when driven by the Tomato -yellow-leaf-curl-virus (TYLCV) -derived universal vector IL-60.Petroleum-derived plastics are not degradable, and are therefore harmful to the environment. Fermentation of bacteria
Y S Liang et al.
Journal of animal science, 95(4), 1727-1738 (2017-05-04)
The objective of this study was to evaluate the effect of residual feed intake (RFI) on rumen function in finishing lambs. A total of 60 male Hu lambs (average initial BW = 25.2 ± 2.5kg) were used and were offered
Andres García et al.
Microbial cell factories, 17(1), 10-10 (2018-01-24)
Azotobacter vinelandii is a bacterium that produces alginate and polyhydroxybutyrate (P3HB); however, the role of NAD(P)H/NAD(P) The results reveal that changes in the oxygen availability have an important effect on the metabolic fluxes and intracellular NADPH/NADP We have demonstrated that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service