Skip to Content
Merck
All Photos(6)

Key Documents

100536

Sigma-Aldrich

4-Aminobenzoic acid

99%, for peptide synthesis, ReagentPlus®

Synonym(s):

PABA, Vitamin Bx, Vitamin H1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CO2H
CAS Number:
Molecular Weight:
137.14
Beilstein:
471605
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

4-Aminobenzoic acid, ReagentPlus®, 99%

Quality Level

product line

ReagentPlus®

Assay

99%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

187-189 °C (lit.)

solubility

95% ethanol: soluble 5%, clear to slightly hazy, colorless to yellow

density

1.374 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

Nc1ccc(cc1)C(O)=O

InChI

1S/C7H7NO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,8H2,(H,9,10)

InChI key

ALYNCZNDIQEVRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Aminobenzoic acid (4-ABA) acts as an bacterial cofactor and participates in the synthesis of folic acid.[1] It is a versatile reagent for structure extension through linear hydrogen-bonding associations, involving both the carboxylic acid and amine functional groups.[1] Formation of 4-ABA monolayer film on glassy carbon electrode (GCE) by amino cation radical method has been described.[2] Molecular adducts of 4-ABA have been prepared and their characterization by infrared spectroscopy and X-ray diffration methods have been reported.[3]

Application

4-Aminobenzoic acid may be used in the following studies:
  • Covalent modification of glassy carbon electrode.[4]
  • Synthesis of 1,4-(2-chlorobenzylidene)aminobenzoic acid (Schiff base).[5]
  • Spectrophotometric determination of the trace amounts of dopamine, methyldopa and levodopa.[6]
  • Synthesis of 1:1 proton-transfer compound with 5-sulfosalicylic acid.[1]

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

339.8 °F - closed cup

Flash Point(C)

171 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The 1: 1 proton-transfer compound of 5-sulfosalicylic acid with 4-aminobenzoic acid.
Smith G, et al.
Acta Crystallographica Section E, Structure Reports Online, 61(2), 313-316 (2005)
Covalent modification of a glassy carbon surface by 4-aminobenzoic acid and its application in fabrication of a polyoxometalates-consisting monolayer and multilayer films.
Liu J, et al.
Langmuir, 16(19), 7471-7476 (2000)
Gehad G Mohamed et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 62(4-5), 1140-1150 (2005-06-16)
Metal complexes of Schiff base derived from 2-thiophene carboxaldehyde and 2-aminobenzoic acid (HL) are reported and characterized based on elemental analyses, IR, 1H NMR, solid reflectance, magnetic moment, molar conductance and thermal analysis (TGA). The ligand dissociation as well as
Layer-by-layer assembly of multilayer films consisting of silicotungstate and a cationic redox polymer on 4-aminobenzoic acid modified glassy carbon electrode and their electrocatalytic effects.
Cheng L, et al.
Analytica Chimica Acta, 34(12), 1093-1100 (1999)
Spectrophotometric determination of catecholamines based on their oxidation reaction followed by coupling with 4-aminobenzoic acid.
Madrakian T, et al.
Journal of the Brazilian Chemical Society, 17(7), 1259-1265 (2006)

Questions

  1. Hallo, wie lange ist die Haltbarkeit des Produkts? auf dem Gebinde und dem Zertifikat gibt es dazu keine Angaben.

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Customers should routinely inspect products with no expiration date or recommended retest date to ensure they perform as expected. These products are also subject to a one-year warranty from the date of shipment.

      For more information, you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service